Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00450829_g1
          See other SIT1 GE Assays ›
          Gene Symbol:
          Sit1
          Gene Name:
          suppression inducing transmembrane adaptor 1
          Gene Aliases:
          Sit
          Chromosome Location:
          Chr.4: 43482083 - 43483709 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00450829_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sit1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019436.3 NP_062309.2 4 - 5 437 80
          GenBank mRNA AJ236881.1 - 4 - 5 413 80
          AK079882.1 - 4 - 5 439 80
          BC064055.1 - 4 - 5 450 576098 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002525.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sit1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          signal transduction
          regulation of T cell activation
          kinase binding
          SH2 domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871967 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871971 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871969 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871970 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 BC064055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 NM_019436.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 AJ236881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 AK079882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 BC064055.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 NM_019436.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 AJ236881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144632 AK079882.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894998 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170695 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 BC064055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 NM_019436.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 AJ236881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 AK079882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 BC064055.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 NM_019436.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 AJ236881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144632 AK079882.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 BC064055.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 NM_019436.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 AJ236881.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 AK079882.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 BC064055.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 NM_019436.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 AJ236881.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P144632 AK079882.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22871928 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170695 - -
          Whole Mouse Genome G4122A A_51_P144632 BC064055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 NM_019436.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 AJ236881.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 AK079882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 BC064055.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 NM_019436.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 AJ236881.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144632 AK079882.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136181 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170695 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03900346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912858 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03686538 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04024624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03900344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04024608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03912856 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871971 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871969 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871970 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22871967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22871887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03685056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03335233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03912875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03912862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03302806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03912864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03694117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871900 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22871943 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512483 - -
          Affymetrix HT MG-430 PM Array Plate 1418751_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512480 - -
          Mouse Genome 430 2.0 Array 1418751_PM_at - -
          Mouse Genome 430 2.0 Array 1418751_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54390_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54390_at - -
          Affymetrix Mouse Clariom S Array PSR0400020518.mm.1 TC0400002525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020518.mm.1 NM_019436.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020518.mm.1 NM_019436.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020515.mm.1 TC0400002525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020515.mm.1 NM_019436.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020515.mm.1 NM_019436.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020516.mm.1 TC0400002525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020516.mm.1 NM_019436.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020516.mm.1 NM_019436.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020517.mm.1 TC0400002525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020517.mm.1 NM_019436.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020517.mm.1 NM_019436.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020520.mm.1 TC0400002525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020520.mm.1 NM_019436.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020520.mm.1 NM_019436.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020521.mm.1 TC0400002525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400020521.mm.1 NM_019436.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400020521.mm.1 NM_019436.3 Refseq
          Mouse Expression Set 430 1418751_PM_at - -
          Mouse Expression Set 430 1418751_at - -
          Murine Genome U74v2 Set 104660_at - -
          Murine Genome U74v2 Set 104660_PM_at - -
          Mouse Genome 430A 2.0 Array 1418751_PM_at - -
          Mouse Genome 430A 2.0 Array 1418751_at - -
          Affymetrix HT Mouse Genome 430A Array 1418751_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418751_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 416696 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4200717 - -
          Illumina mouse-6 v1.0 expression beadchip 4200717 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258509 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258509 - -
          mouse-6 v1.1 (Illumina) scl24404.5.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24404.5.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200717 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2890954 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258509 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2890954 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline