Search
Search

Myo10| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019472.2 | NP_062345.2 | 27 - 28 | 4271 | 62 | |
| GenBank mRNA | AJ249706.2 | - | 27 - 28 | 3895 | 62 | |
| AK122371.1 | - | 7 - 8 | 2053 | 62 | ||
| BC024692.1 | - | 3 - 4 | 397 | 62 | ||
Set Membership: |
|