Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00450916_s1
          See other OLFR71 GE Assays ›
          Gene Symbol:
          Olfr71
          Gene Name:
          olfactory receptor 71
          Gene Aliases:
          MOR262-4, mOR17
          Chromosome Location:
          Chr.4: 43705628 - 43706566 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00450916_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019486.1 NP_062359.1 1 450 91
          GenBank mRNA BC100681.1 - 1 449 91
          BC100683.1 - 1 448 91
          BC100684.1 - 1 449 91
          BC100686.1 - 1 449 91
          BC115841.1 - 1 449 91
          BC141882.1 - 1 450 40041946 91
          BC141892.1 - 1 450 40041948 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr71

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053883 BC141882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053883 BC141892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053883 BC100686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053883 BC115841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2053883 NM_019486.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554374 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178657 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053883 BC141882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053883 BC141892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053883 BC100686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053883 BC115841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053883 NM_019486.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326357 BC141882.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326357 BC141892.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326357 BC100686.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326357 BC115841.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P326357 NM_019486.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873336 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178657 - -
          Whole Mouse Genome G4122A A_51_P326357 BC141882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326357 BC141892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326357 BC100686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326357 BC115841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326357 NM_019486.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115037 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178657 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03812716 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03812707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03887249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03814459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03764746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03330435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03756784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03812707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03812718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04024847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03756781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03812709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22873346 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512627 - -
          Affymetrix HT MG-430 PM Array Plate 1422373_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512626 - -
          Mouse Genome 430 2.0 Array 1422373_PM_at - -
          Mouse Genome 430 2.0 Array 1422373_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56015_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56015_PM_at - -
          Mouse Expression Set 430 1422373_PM_at - -
          Mouse Expression Set 430 1422373_at - -
          Murine Genome U74v2 Set 97004_PM_at - -
          Murine Genome U74v2 Set 97004_at - -
          Mouse Genome 430A 2.0 Array 1422373_at - -
          Mouse Genome 430A 2.0 Array 1422373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422373_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422373_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 480210 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4810341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215564 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215564 - -
          mouse-6 v1.1 (Illumina) scl2470.1.1_205-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl2470.1.1_205-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215564 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215564 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline