Search
Search

Vps29| Gene Symbol: Vps29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019780.1 | NP_062754.1 | 2 - 3 | 253 | 73 | |
| GenBank mRNA | AF193794.1 | - | 2 - 3 | 253 | 73 | |
| AK004103.1 | - | 3 - 4 | 270 | 73 | ||
| AK036426.1 | - | 2 - 3 | 241 | 73 | ||
| AK150330.1 | - | 2 - 3 | 263 | 73 | ||
| BC005663.1 | - | 2 - 3 | 242 | 2645432 | 73 | |
Set Membership: |
|