Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451559_s1
          See other OLFR140 GE Assays ›
          Gene Symbol:
          Olfr140
          Gene Name:
          olfactory receptor 140
          Gene Aliases:
          A16, MOR235-1, Ora16
          Chromosome Location:
          Chr.2: 90051414 - 90063591 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451559_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr140
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020515.1 NP_065261.1 1 50 98
          GenBank mRNA BC111111.1 - 1 47 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004077.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr140

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          transmembrane signaling receptor activity
          G-protein coupled receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407811 BC111111.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407811 BC141888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407811 NM_020515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407811 BC111111.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407811 BC141888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P407811 NM_020515.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169016 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407811 BC111111.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407811 BC141888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407811 NM_020515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407811 BC111111.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407811 BC141888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407811 NM_020515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407811 BC111111.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407811 BC141888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407811 NM_020515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407811 BC111111.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407811 BC141888.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P407811 NM_020515.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169016 - -
          Whole Mouse Genome G4122A A_51_P407811 BC111111.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407811 BC141888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407811 NM_020515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407811 BC111111.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407811 BC141888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P407811 NM_020515.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113321 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02216625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01610589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01412662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415713 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484862 - -
          Affymetrix HT MG-430 PM Array Plate 1450604_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10484861 - -
          Mouse Genome 430 2.0 Array 1450604_PM_at - -
          Mouse Genome 430 2.0 Array 1450604_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57272_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57272_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200034117.mm.1 NM_020515.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034117.mm.1 NM_020515.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200034117.mm.1 TC0200004077.mm.1 MTA TC
          Mouse Expression Set 430 1450604_at - -
          Mouse Expression Set 430 1450604_PM_at - -
          Murine Genome U74v2 Set 96986_at - -
          Murine Genome U74v2 Set 96986_PM_at - -
          Mouse Genome 430A 2.0 Array 1450604_at - -
          Mouse Genome 430A 2.0 Array 1450604_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450604_at - -
          Affymetrix HT Mouse Genome 430A Array 1450604_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 483196 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1980025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232633 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232633 - -
          mouse-6 v1.1 (Illumina) scl4266.1.1_271-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4266.1.1_271-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3000343 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3000343 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline