Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451600_g1
          See other PTH GE Assays ›
          Gene Symbol:
          Pth
          Gene Name:
          parathyroid hormone
          Gene Aliases:
          Pthp
          Chromosome Location:
          Chr.7: 113385576 - 113388573 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451600_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pth
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020623.2 NP_065648.1 2 - 3 211 82
          GenBank mRNA AK138677.1 - 2 - 3 209 82
          BC099456.1 - 2 - 3 196 30845904 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004128.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pth

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          skeletal system development
          transcription from RNA polymerase II promoter
          cellular calcium ion homeostasis
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          Rho protein signal transduction
          cell-cell signaling
          positive regulation of signal transduction
          regulation of gene expression
          positive regulation of bone mineralization
          positive regulation of cAMP biosynthetic process
          cellular macromolecule biosynthetic process
          bone resorption
          positive regulation of glycogen biosynthetic process
          positive regulation of ossification
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of glucose import
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          hormone activity
          parathyroid hormone receptor binding
          type 1 parathyroid hormone receptor binding
          peptide hormone receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230496 BC099456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230496 NM_020623.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230496 AK138677.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230496 BC099456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230496 NM_020623.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P230496 AK138677.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 354493 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136439 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230496 BC099456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230496 NM_020623.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230496 AK138677.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230496 BC099456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230496 NM_020623.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P230496 AK138677.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230496 BC099456.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230496 NM_020623.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230496 AK138677.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230496 BC099456.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230496 NM_020623.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230496 AK138677.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25430313 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136439 - -
          Whole Mouse Genome G4122A A_51_P230496 BC099456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230496 NM_020623.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230496 AK138677.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230496 BC099456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230496 NM_020623.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230496 AK138677.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111391 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06298616 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06298627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06124744 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06594435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06298614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25430265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06117957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06240191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06545836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06124752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06001473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25430328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10567036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567037 - -
          Affymetrix HT MG-430 PM Array Plate 1419345_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567034 - -
          Mouse Genome 430 2.0 Array 1419345_PM_at - -
          Mouse Genome 430 2.0 Array 1419345_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19226_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19226_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700034214.mm.1 NM_020623.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034214.mm.1 NM_020623.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034214.mm.1 TC0700004128.mm.1 MTA TC
          Mouse Expression Set 430 1419345_PM_at - -
          Mouse Expression Set 430 1419345_at - -
          Murine Genome U74v2 Set 102078_at - -
          Murine Genome U74v2 Set 102078_PM_at - -
          Mouse Genome 430A 2.0 Array 1419345_at - -
          Mouse Genome 430A 2.0 Array 1419345_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419345_at - -
          Affymetrix HT Mouse Genome 430A Array 1419345_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 831949 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6650706 - -
          Illumina mouse-6 v1.0 expression beadchip 6650706 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006650706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253980 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253980 - -
          mouse-6 v1.1 (Illumina) scl30791.3.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 6650706 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30791.3.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6650706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2786030 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2786030 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline