Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452625_m1
          See other MRPL11 GE Assays ›
          Gene Symbol:
          Mrpl11
          Gene Name:
          mitochondrial ribosomal protein L11
          Gene Aliases:
          2410001P07Rik, L11mt
          Chromosome Location:
          Chr.19: 4962152 - 4966995 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452625_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025553.4 NP_079829.1 4 - 5 560 98
          GenBank mRNA AB049639.1 - 4 - 5 538 98
          AK002797.1 - 4 - 5 515 98
          AK010329.1 - 4 - 5 552 98
          AK167247.1 - 4 - 5 528 98
          BC046750.1 - 4 - 5 513 5692108 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000050.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl11

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926078 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434240 AK010329.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434240 AK010329.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77687 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154906 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434240 AK010329.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434240 AK010329.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434240 AK010329.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434240 AK010329.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926071 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154906 - -
          Whole Mouse Genome G4122A A_51_P434240 AK010329.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434240 AK010329.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123674 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115286 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14338305 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14338331 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14338337 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14338304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14338310 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31926078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14338336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14173244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14338335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14338316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14396305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14434764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14338340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926090 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460494 - -
          Affymetrix HT MG-430 PM Array Plate 1437131_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1437839_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1417918_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438080_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460492 - -
          Mouse Genome 430 2.0 Array 1437131_x_at - -
          Mouse Genome 430 2.0 Array 1438080_at - -
          Mouse Genome 430 2.0 Array 1438080_PM_at - -
          Mouse Genome 430 2.0 Array 1437839_PM_x_at - -
          Mouse Genome 430 2.0 Array 1417918_PM_at - -
          Mouse Genome 430 2.0 Array 1417918_at - -
          Mouse Genome 430 2.0 Array 1437131_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437839_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66419_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66419_at - -
          Affymetrix Mouse Clariom S Array PSR1900000675.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000675.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000675.mm.1 TC1900000050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000671.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000671.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000671.mm.1 TC1900000050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000677.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000677.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000677.mm.1 TC1900000050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000673.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000673.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000673.mm.1 TC1900000050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000674.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000674.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000674.mm.1 TC1900000050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000676.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000676.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000676.mm.1 TC1900000050.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000672.mm.1 NM_025553.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000672.mm.1 NM_025553.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000672.mm.1 TC1900000050.mm.1 MTA TC
          Mouse Expression Set 430 1417918_at - -
          Mouse Expression Set 430 1437839_PM_x_at - -
          Mouse Expression Set 430 1417918_PM_at - -
          Mouse Expression Set 430 1437131_PM_x_at - -
          Mouse Expression Set 430 1438080_at - -
          Mouse Expression Set 430 1438080_PM_at - -
          Mouse Expression Set 430 1437839_x_at - -
          Mouse Expression Set 430 1437131_x_at - -
          Murine Genome U74v2 Set 98876_at - -
          Murine Genome U74v2 Set 98876_PM_at - -
          Mouse Genome 430A 2.0 Array 1438080_at - -
          Mouse Genome 430A 2.0 Array 1417918_PM_at - -
          Mouse Genome 430A 2.0 Array 1438080_PM_at - -
          Mouse Genome 430A 2.0 Array 1437131_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1417918_at - -
          Mouse Genome 430A 2.0 Array 1437839_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437131_x_at - -
          Mouse Genome 430A 2.0 Array 1437839_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437839_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1417918_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437131_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437839_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438080_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438080_at - -
          Affymetrix HT Mouse Genome 430A Array 1417918_at - -
          Affymetrix HT Mouse Genome 430A Array 1437131_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913623 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 110215 - -
          Illumina mouse-6 v1.0 expression beadchip 110215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232776 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232776 - -
          mouse-6 v1.1 (Illumina) scl52826.3.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 110215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52826.3.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232776 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232776 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52826.3.1_11-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline