Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452627_m1
          See other POLR2E GE Assays ›
          Gene Symbol:
          Polr2e
          Gene Name:
          polymerase (RNA) II (DNA directed) polypeptide E
          Gene Aliases:
          2410021N14Rik, 25kDa, AW208866, RPB5, XAP4
          Chromosome Location:
          Chr.10: 80035953 - 80039659 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452627_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Polr2e
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025554.2 NP_079830.2 2 - 3 251 61
          GenBank mRNA AK010567.1 - 2 - 3 259 61
          AK015823.1 - 2 - 3 249 61
          AK088610.1 - 2 - 3 308 61
          AK132109.1 - 2 - 3 306 61
          AK144668.1 - 2 - 3 312 61
          AK145201.1 - 2 - 3 315 61
          BC005599.1 - 2 - 3 303 61
          BC026842.1 - 2 - 3 242 4210827 61
          BC045521.1 - 2 - 3 251 2647754 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002488.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-directed RNA polymerase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          transcription from RNA polymerase II promoter
          RNA polymerase I activity
          RNA polymerase II activity
          RNA polymerase III activity
          DNA binding
          DNA-directed RNA polymerase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280575 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280577 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 BC045521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 AK144668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 AK145201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 BC005599.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 BC026842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 BC045521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 AK144668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 AK145201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 BC005599.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P362033 BC026842.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119680 BC037681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P119680 AK015823.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38830 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 BC045521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 AK144668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 AK145201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 BC005599.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 BC026842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 BC045521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 AK144668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 AK145201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 BC005599.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P362033 BC026842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119680 BC037681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P119680 AK015823.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239456 AK010567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 BC045521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 AK144668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 AK145201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 BC005599.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 BC026842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 BC045521.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 AK144668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 AK145201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 BC005599.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362033 BC026842.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239456 AK010567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P973604 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08968896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08968895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280577 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27280575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27280569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08942741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08495371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08804970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08495374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08612484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08968893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08850658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08889096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08556712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08556713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08595508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08434831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280593 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08320537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08595513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08320536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27280536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370719 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370712 - -
          Affymetrix HT MG-430 PM Array Plate 1458326_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417138_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1451093_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370708 - -
          Mouse Genome 430 2.0 Array 1417138_PM_s_at - -
          Mouse Genome 430 2.0 Array 1458326_at - -
          Mouse Genome 430 2.0 Array 1417138_s_at - -
          Mouse Genome 430 2.0 Array 1451093_at - -
          Mouse Genome 430 2.0 Array 1458326_PM_at - -
          Mouse Genome 430 2.0 Array 1451093_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66420_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66420_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000018281.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018281.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018281.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018284.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018279.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018279.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018279.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018278.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018278.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018278.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018290.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018290.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018290.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018283.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018283.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018283.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018282.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018291.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018291.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018291.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018285.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018285.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018285.mm.1 TC1000002488.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018288.mm.1 NM_025554.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018288.mm.1 NM_025554.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018288.mm.1 TC1000002488.mm.1 MTA TC
          Mouse Expression Set 430 1417138_PM_s_at - -
          Mouse Expression Set 430 1451093_at - -
          Mouse Expression Set 430 1451093_PM_at - -
          Mouse Expression Set 430 1417138_s_at - -
          Mouse Expression Set 430 1458326_PM_at - -
          Mouse Expression Set 430 1458326_at - -
          Murine Genome U74v2 Set 93325_at - -
          Murine Genome U74v2 Set 93325_PM_at - -
          Murine Genome U74v2 Set 165866_PM_f_at - -
          Murine Genome U74v2 Set 165866_f_at - -
          Mouse Genome 430A 2.0 Array 1417138_s_at - -
          Mouse Genome 430A 2.0 Array 1451093_at - -
          Mouse Genome 430A 2.0 Array 1451093_PM_at - -
          Mouse Genome 430A 2.0 Array 1417138_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451093_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417138_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417138_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451093_at - -
          Affymetrix HT Mouse Genome 430B Array 1458326_at - -
          Affymetrix HT Mouse Genome 430B Array 1458326_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 905496 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218675 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234905 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218675 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234905 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2709446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234905 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline