Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452668_m1
          See other POLE4 GE Assays ›
          Gene Symbol:
          Pole4
          Gene Name:
          polymerase (DNA-directed), epsilon 4 (p12 subunit)
          Gene Aliases:
          2400007P05Rik, 5830430F06Rik
          Chromosome Location:
          Chr.6: 82618925 - 82652936 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452668_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pole4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025882.3 NP_080158.1 1 - 2 248 75
          GenBank mRNA AK010135.1 - 1 - 2 243 75
          AK010300.1 - 1 - 2 249 75
          AK011595.1 - 1 - 2 260 75
          AK145365.1 - 1 - 2 254 75
          AK160582.1 - 1 - 2 260 75
          AK167631.1 - 1 - 2 238 75
          BC023189.1 - 1 - 2 226 4952949 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002548.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pole4

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          histone H3 acetylation
          molecular_function
          DNA binding
          DNA-directed DNA polymerase activity
          transferase activity
          nucleotidyltransferase activity
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589126 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589129 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450924 AK017956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450924 AK017956.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 165378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365273 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22264 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109349 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156947 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450924 AK017956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450924 AK017956.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P465043 AK167631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P465043 BC023189.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P465043 NM_025882.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P465043 AK010135.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P465043 AK010300.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450924 AK017956.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P450924 AK017956.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24589145 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05959485 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05959491 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156947 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05959499 - -
          Whole Mouse Genome G4122A A_52_P335478 AK167631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335478 AK182530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335478 AK160582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335478 BC023189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335478 NM_025882.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465043 AK167631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465043 BC023189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465043 NM_025882.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465043 AK010135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P465043 AK010300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450924 AK017956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450924 AK017956.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102075 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156947 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05959499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05847047 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05847051 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589131 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589125 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589123 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589129 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589127 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24589126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24589080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05847041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05607854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05191824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05669324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05240752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05847044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05607856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05674474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05826414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05607858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05516351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24589046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545587 - -
          Affymetrix HT MG-430 PM Array Plate 1423372_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432286_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423371_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545583 - -
          Mouse Genome 430 2.0 Array 1423371_PM_at - -
          Mouse Genome 430 2.0 Array 1423371_at - -
          Mouse Genome 430 2.0 Array 1432286_PM_at - -
          Mouse Genome 430 2.0 Array 1423372_at - -
          Mouse Genome 430 2.0 Array 1423372_PM_at - -
          Mouse Genome 430 2.0 Array 1432286_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66979_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66979_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600019691.mm.1 TC0600002548.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019691.mm.1 NM_025882.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019691.mm.1 NM_025882.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019695.mm.1 TC0600002548.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600019695.mm.1 NM_025882.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019695.mm.1 NM_025882.3 Refseq
          Mouse Expression Set 430 1423371_at - -
          Mouse Expression Set 430 1423372_at - -
          Mouse Expression Set 430 1423372_PM_at - -
          Mouse Expression Set 430 1423371_PM_at - -
          Murine Genome U74v2 Set 95121_at - -
          Murine Genome U74v2 Set 95122_g_at - -
          Murine Genome U74v2 Set 95122_PM_g_at - -
          Murine Genome U74v2 Set 95121_PM_at - -
          Mouse Genome 430A 2.0 Array 1423372_PM_at - -
          Mouse Genome 430A 2.0 Array 1423371_at - -
          Mouse Genome 430A 2.0 Array 1423371_PM_at - -
          Mouse Genome 430A 2.0 Array 1423372_at - -
          Affymetrix HT Mouse Genome 430A Array 1423371_at - -
          Affymetrix HT Mouse Genome 430A Array 1423372_at - -
          Affymetrix HT Mouse Genome 430A Array 1423372_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423371_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432286_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432286_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 508321 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6860097 - -
          Illumina mouse-6 v1.0 expression beadchip 6860097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236601 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236601 - -
          mouse-6 v1.1 (Illumina) scl066979.1_124-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066979.1_124-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236601 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236601 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl066979.1_124-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0