| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700009739.mm.1 |
NM_026498.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009739.mm.1 |
NM_026498.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009739.mm.1 |
TC0700001110.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009745.mm.1 |
NM_026498.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009745.mm.1 |
NM_026498.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009745.mm.1 |
TC0700001110.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009748.mm.1 |
NM_026498.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009748.mm.1 |
NM_026498.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009748.mm.1 |
TC0700001110.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009746.mm.1 |
NM_026498.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009746.mm.1 |
NM_026498.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009746.mm.1 |
TC0700001110.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009740.mm.1 |
NM_026498.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009740.mm.1 |
NM_026498.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009740.mm.1 |
TC0700001110.mm.1 |
MTA TC |