Search
Search

Pdgfrl| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026840.3 | NP_081116.3 | 2 - 3 | 494 | 64 | |
| GenBank mRNA | AK004179.1 | - | 2 - 3 | 386 | 64 | |
| AK132625.1 | - | 2 - 3 | 495 | 64 | ||
| BC058275.1 | - | 2 - 3 | 486 | 6515674 | 64 | |
Set Membership: |
|