Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00452999_s1
          See other STARD6 GE Assays ›
          Gene Symbol:
          Stard6
          Gene Name:
          StAR-related lipid transfer (START) domain containing 6
          Gene Aliases:
          1700011K09Rik, 4833424I06Rik, 4933429L05Rik, AI644424
          Chromosome Location:
          Chr.18: 70472454 - 70505254 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00452999_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stard6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289648.1 NP_001276577.1 9 827 123
          NM_001289649.1 NP_001276578.1 9 806 123
          NM_001289650.1 NP_001276579.1 9 841 123
          NM_029019.4 NP_083295.1 9 896 123
          GenBank mRNA AF480303.1 - 6 543 123
          AK005873.1 - 9 841 123
          AK014759.1 - 9 807 123
          AK016984.1 - 9 807 123
          BC061022.1 - 7 579 6774873 123
          BC087893.1 - 9 818 6774832 123
          JF440953.1 - 7 615 123
          JF440954.1 - 7 615 123

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000758.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stard6

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          lipid transport
          biological_process
          lipid binding
          cholesterol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31809752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31809751 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974088 AK005873.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974088 AK014759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974088 BC061022.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974088 BC087893.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974088 NM_029019.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85765 AK014759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85765 NM_029019.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85765 AK014759.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85765 NM_029019.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285417 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110530 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165155 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161092 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85765 AK014759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85765 NM_029019.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85765 AK014759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85765 NM_029019.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974088 AK005873.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974088 AK014759.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974088 BC061022.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974088 BC087893.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974088 NM_029019.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31809772 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09043851 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09043863 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165155 - -
          Whole Mouse Genome G4122A A_52_P85765 AK014759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85765 NM_029019.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85765 AK014759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85765 NM_029019.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125391 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136883 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165155 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13794757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13794763 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13794754 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13794753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13794758 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31809751 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31809752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31809839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14011789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13705360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14071708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13794758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13759662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14011790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31809797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13751286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10456559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456557 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363184 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363183 - -
          Affymetrix Mouse Gene 1.0 ST Array 10363182 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456560 - -
          Affymetrix HT MG-430 PM Array Plate 1426317_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10456556 - -
          Affymetrix Mouse Gene 1.1 ST Array 10363181 - -
          Mouse Genome 430 2.0 Array 1426317_at - -
          Mouse Genome 430 2.0 Array 1426317_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800005508.mm.1 NM_001289648.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005508.mm.1 NM_001289649.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005508.mm.1 NM_001289650.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005508.mm.1 NM_029019.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005508.mm.1 TC1800000758.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005506.mm.1 NM_001289648.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005506.mm.1 NM_001289649.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005506.mm.1 NM_001289650.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005506.mm.1 NM_029019.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005506.mm.1 TC1800000758.mm.1 MTA TC
          Mouse Expression Set 430 1426317_at - -
          Mouse Expression Set 430 1426317_PM_at - -
          Mouse Genome 430A 2.0 Array 1426317_at - -
          Mouse Genome 430A 2.0 Array 1426317_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426317_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426317_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 816158 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940541 - -
          Illumina mouse-6 v1.0 expression beadchip 6940541 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940541 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215885 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215885 - -
          mouse-6 v1.1 (Illumina) scl51791.11_399-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940541 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51791.11_399-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215885 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215885 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline