Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00453250_m1
          See other GSDMC GE Assays ›
          Gene Symbol:
          Gsdmc
          Gene Name:
          gasdermin C
          Gene Aliases:
          Gsdmc1, Mlze
          Chromosome Location:
          Chr.15: 63775971 - 63808739 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00453250_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsdmc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031378.3 NP_113555.1 4 - 5 827 68
          GenBank mRNA AB042406.1 - 3 - 4 724 68
          AK137662.1 - 4 - 5 828 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002343.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gsdmc

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150489 AK137662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150489 NM_031378.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150489 AB042406.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150489 AK137662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150489 NM_031378.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P150489 AB042406.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508895 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243893 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5125 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152448 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150489 AK137662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150489 NM_031378.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150489 AB042406.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150489 AK137662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150489 NM_031378.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P150489 AB042406.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03053106 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08393291 - -
          Whole Mouse Genome G4122A A_51_P150489 AK137662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150489 NM_031378.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150489 AB042406.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150489 AK137662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150489 NM_031378.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P150489 AB042406.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106103 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127060 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30277454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11989085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12396590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12436195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11975415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12194415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11951524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30277575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10428945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428951 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428944 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428953 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428954 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428962 - -
          Affymetrix HT MG-430 PM Array Plate 1419591_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10428943 - -
          Affymetrix Mouse Gene 1.1 ST Array 10428960 - -
          Mouse Genome 430 2.0 Array 1419591_at - -
          Mouse Genome 430 2.0 Array 1419591_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83492_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83492_at - -
          Affymetrix Mouse Clariom S Array PSR1500012106.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012106.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012106.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012107.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012107.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012107.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012108.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012108.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012108.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012109.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012109.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012109.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012110.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012110.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012110.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012111.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012111.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012111.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012112.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012112.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012112.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012114.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012114.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012114.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500012113.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012113.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500012113.mm.1 TC1500002343.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500019350.mm.1 NM_031378.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019350.mm.1 NM_031378.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500019350.mm.1 TC1500002343.mm.1 MTA TC
          Mouse Expression Set 430 1419591_at - -
          Mouse Expression Set 430 1419591_PM_at - -
          Mouse Genome 430A 2.0 Array 1419591_at - -
          Mouse Genome 430A 2.0 Array 1419591_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419591_at - -
          Affymetrix HT Mouse Genome 430A Array 1419591_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 648294 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6450450 - -
          MouseWG-6 v2 Expression BeadChip 5290411 - -
          MouseWG-6 v2 Expression BeadChip 106510451 - -
          Illumina mouse-6 v1.0 expression beadchip 106510451 - -
          Illumina mouse-6 v1.0 expression beadchip 6450450 - -
          Illumina mouse-6 v1.0 expression beadchip 7040451 - -
          Illumina mouse-6 v1.0 expression beadchip 5290411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005290411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240230 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257797 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2678631 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257797 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256484 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2678631 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240230 - -
          mouse-6 v1.1 (Illumina) scl083492.1_0-S - -
          mouse-6 v1.1 (Illumina) scl0083492.2_313-S - -
          mouse-6 v1.1 (Illumina) GI_13878198-S - -
          mouse-6 v1.1 (Illumina) scl0083492.2_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 5290411 - -
          Illumina Sentrix mouse-6.v1 bead array 6450450 - -
          Illumina Sentrix mouse-6.v1 bead array 106510451 - -
          Illumina Sentrix mouse-6.v1 bead array 7040451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl083492.1_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0083492.2_75-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0083492.2_313-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106510451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5290411 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2730216 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240230 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257797 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2730216 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240230 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257797 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0083492.2_75-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline