Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00453413_s1
          See other FERD3L GE Assays ›
          Gene Symbol:
          Ferd3l
          Gene Name:
          Fer3-like (Drosophila)
          Gene Aliases:
          Mnato3, N-twist, Nato, Nato3, Ntwist, bHLHa31, fer3
          Chromosome Location:
          Chr.12: 33928425 - 33929310 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00453413_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ferd3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033522.2 NP_277057.1 1 109 62
          GenBank mRNA AF517121.1 - 1 109 62
          BC111576.1 - 1 101 40039965 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000296.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ferd3l

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          floor plate development
          negative regulation of transcription, DNA-templated
          cell development
          regulation of neurogenesis
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977220 NM_033522.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977220 AF517121.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 889036 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156409 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977220 NM_033522.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977220 AF517121.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28344165 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156409 - -
          Whole Mouse Genome G4122A A_51_P276753 NM_033522.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276753 AF517121.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276753 BC111576.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116137 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123180 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156409 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10132286 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10132297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10132291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10132283 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10132287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10132292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28344133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10099577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10435745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10099573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10374799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10293512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10102140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09943254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10374795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10371741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09842153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28344173 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395318 - -
          Affymetrix HT MG-430 PM Array Plate 1422410_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395317 - -
          Mouse Genome 430 2.0 Array 1422410_PM_at - -
          Mouse Genome 430 2.0 Array 1422410_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114712_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114712_at - -
          Affymetrix Mouse Clariom S Array PSR1200002284.mm.1 NM_033522.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002284.mm.1 NM_033522.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002284.mm.1 TC1200000296.mm.1 MTA TC
          Mouse Expression Set 430 1422410_at - -
          Mouse Expression Set 430 1422410_PM_at - -
          Mouse Genome 430A 2.0 Array 1422410_PM_at - -
          Mouse Genome 430A 2.0 Array 1422410_at - -
          Affymetrix HT Mouse Genome 430A Array 1422410_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422410_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 856770 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3850673 - -
          Illumina mouse-6 v1.0 expression beadchip 3850673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2724181 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2724181 - -
          mouse-6 v1.1 (Illumina) scl43292.1.1_224-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43292.1.1_224-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2724181 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2724181 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline