Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454186_s1
          See other VMN1R234 GE Assays ›
          Gene Symbol:
          Vmn1r234
          Gene Name:
          vomeronasal 1 receptor 234
          Gene Aliases:
          V1rf1
          Chromosome Location:
          Chr.17: 21228826 - 21229815 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454186_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r234
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134198.1 NP_598959.1 1 55 77
          GenBank mRNA BC127026.1 - 1 55 40044289 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000264.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r234

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082539 NM_134198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082539 BC127026.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131294 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082539 NM_134198.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082539 BC127026.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100218 NM_134198.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100218 BC127026.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085479 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131294 - -
          Whole Mouse Genome G4122A A_51_P100218 NM_134198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P100218 BC127026.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101172 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131294 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457183 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457175 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13279981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442183 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442182 - -
          Affymetrix Mouse Clariom S Array PSR1700001953.mm.1 NM_134198.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001953.mm.1 NM_134198.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001953.mm.1 TC1700000264.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 540921 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3990181 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990181 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242477 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242477 - -
          mouse-6 v1.1 (Illumina) scl8630.1.1_120-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990181 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8630.1.1_120-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990181 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242477 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242477 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline