Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454196_s1
          See other VMN1R235 GE Assays ›
          Gene Symbol:
          Vmn1r235
          Gene Name:
          vomeronasal 1 receptor 235
          Gene Aliases:
          V1rf2
          Chromosome Location:
          Chr.17: 21260427 - 21262863 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454196_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r235
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134199.3 NP_598960.3 2 383 107
          GenBank mRNA AF394953.1 - 2 384 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000266.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r235

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P495426 AF394953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P495426 NM_134199.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P495426 AF394953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P495426 NM_134199.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130855 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167948 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P495426 AF394953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P495426 NM_134199.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P495426 AF394953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P495426 NM_134199.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495426 AF394953.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495426 NM_134199.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495426 AF394953.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P495426 NM_134199.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085552 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167948 - -
          Whole Mouse Genome G4122A A_51_P495426 AF394953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495426 NM_134199.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495426 AF394953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P495426 NM_134199.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104720 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13271702 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13271688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13271695 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13271684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13271696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442185 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442186 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442184 - -
          Affymetrix Mouse Clariom S Array PSR1700001956.mm.1 NM_134199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001956.mm.1 NM_134199.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001956.mm.1 TC1700000266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001955.mm.1 NM_134199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001955.mm.1 NM_134199.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001955.mm.1 TC1700000266.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 789038 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4120537 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120537 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2486295 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2486295 - -
          mouse-6 v1.1 (Illumina) scl51062.2.43_40-S - -
          Illumina Sentrix mouse-6.v1 bead array 4120537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51062.2.43_40-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2486295 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2486295 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline