Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454213_s1
          See other VMN1R237 GE Assays ›
          Gene Symbol:
          Vmn1r237
          Gene Name:
          vomeronasal 1 receptor 237
          Gene Aliases:
          V1rf3, Vfrf3
          Chromosome Location:
          Chr.17: 21314017 - 21314886 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454213_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r237
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134200.1 NP_598961.1 1 748 100
          GenBank mRNA BC125543.1 - 1 802 40130058 100
          BC138066.1 - 1 802 8861085 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000268.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r237

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082549 NM_134200.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082549 BC125543.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082549 BC138066.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737849 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166869 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082549 NM_134200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082549 BC125543.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082549 BC138066.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283384 NM_134200.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283384 BC125543.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P283384 BC138066.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085730 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166869 - -
          Whole Mouse Genome G4122A A_51_P283384 NM_134200.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283384 BC125543.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P283384 BC138066.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116491 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201442 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201443 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13201425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13201425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13589900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13239573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13589896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13201427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085746 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442193 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442192 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442191 - -
          Affymetrix Mouse Clariom S Array PSR1700001959.mm.1 NM_134200.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001959.mm.1 NM_134200.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001959.mm.1 TC1700000268.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001958.mm.1 TC1700000268.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 792328 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3060136 - -
          Illumina mouse-6 v1.0 expression beadchip 3060136 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2507665 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2507665 - -
          mouse-6 v1.1 (Illumina) scl8628.1.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8628.1.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2507665 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2507665 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl8628.1.1_3-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline