Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454219_s1
          See other VMN1R236 GE Assays ›
          Gene Symbol:
          Vmn1r236
          Gene Name:
          vomeronasal 1 receptor 236
          Gene Aliases:
          V1rf4
          Chromosome Location:
          Chr.17: 21286533 - 21287653 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454219_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r236
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134201.2 NP_598962.2 1 528 88
          GenBank mRNA BC125459.1 - 1 528 40129974 88
          BC125461.1 - 1 528 40129976 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000267.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r236

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082554 BC125459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082554 BC125461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082554 NM_134201.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28249 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153889 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082554 BC125459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082554 BC125461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082554 NM_134201.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433287 BC125459.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433287 BC125461.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P433287 NM_134201.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31085681 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153889 - -
          Whole Mouse Genome G4122A A_51_P433287 BC125459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433287 BC125461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P433287 NM_134201.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122524 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153889 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13372621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13372618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13372617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13372628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31085676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13372620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13443276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13105806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13372625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31085688 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442190 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442187 - -
          Affymetrix Mouse Clariom S Array PSR1700001957.mm.1 NM_134201.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001957.mm.1 NM_134201.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001957.mm.1 TC1700000267.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2650022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2458586 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2458586 - -
          mouse-6 v1.1 (Illumina) scl8629.1.1_247-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8629.1.1_247-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2458586 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2458586 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline