Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454302_s1
          See other VMN1R81 GE Assays ›
          Gene Symbol:
          Vmn1r81
          Gene Name:
          vomeronasal 1 receptor 81
          Gene Aliases:
          V1rg9
          Chromosome Location:
          Chr.7: 12259759 - 12260679 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454302_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r81
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134210.1 NP_598971.1 1 728 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r81

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24971472 - -
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P115229 NM_134210.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06568393 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06568396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24971496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06568396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24971465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06308721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560018 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560017 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 366083 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2690575 - -
          Illumina mouse-6 v1.0 expression beadchip 2690575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002690575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220012 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220012 - -
          mouse-6 v1.1 (Illumina) scl9204.1.1_223-S - -
          Illumina Sentrix mouse-6.v1 bead array 2690575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9204.1.1_223-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220012 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220012 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline