Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454439_s1
          See other VMN1R67 GE Assays ›
          Gene Symbol:
          Vmn1r67
          Gene Name:
          vomeronasal 1 receptor 67
          Gene Aliases:
          V1re10
          Chromosome Location:
          Chr.7: 10446789 - 10447787 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454439_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134229.2 NP_598990.2 1 753 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r67

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P398089 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P398089 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24966070 - -
          Whole Mouse Genome G4122A A_51_P398089 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06194798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06194805 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06194792 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06194786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24966115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06168864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06129406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06260364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06177964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06290182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06194789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24966054 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549946 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549945 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6900082 - -
          Illumina mouse-6 v1.0 expression beadchip 6900082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006900082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248701 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248701 - -
          mouse-6 v1.1 (Illumina) scl9726.1.1_124-S - -
          Illumina Sentrix mouse-6.v1 bead array 6900082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9726.1.1_124-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2780589 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248701 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2780589 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0