Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00454448_s1
          See other VMN1R66 GE Assays ›
          Gene Symbol:
          Vmn1r66
          Gene Name:
          vomeronasal 1 receptor 66
          Gene Aliases:
          S67, V1re11, V1re3
          Chromosome Location:
          Chr.7: 10273828 - 10275351 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00454448_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134230.3 NP_598991.2 1 960 110
          GenBank mRNA AF394952.1 - 1 959 110
          BC109341.1 - 1 813 40052743 110
          BC109342.1 - 1 813 40052744 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002308.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r66

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 BC109342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 AF394952.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 BC109341.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 NM_134230.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 BC109342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 AF394952.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 BC109341.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P203277 NM_134230.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78546 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159728 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 BC109342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 AF394952.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 BC109341.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 NM_134230.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 BC109342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 AF394952.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 BC109341.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P203277 NM_134230.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 BC109342.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 AF394952.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 BC109341.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 NM_134230.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 BC109342.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 AF394952.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 BC109341.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P203277 NM_134230.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24965549 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159728 - -
          Whole Mouse Genome G4122A A_51_P203277 BC109342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 AF394952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 BC109341.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 NM_134230.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 BC109342.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 AF394952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 BC109341.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P203277 NM_134230.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126174 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06449618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06511613 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06304934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06449621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06511622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06449622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06511612 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06449613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24965563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06304928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06595159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06386481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06304931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06595157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24965574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06205855 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559951 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559948 - -
          Affymetrix Mouse Gene 1.1 ST Array 10559946 - -
          Affymetrix Mouse Clariom S Array PSR0700020705.mm.1 NM_134230.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700020705.mm.1 NM_134230.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700020705.mm.1 TC0700002308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700020706.mm.1 NM_134230.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700020706.mm.1 NM_134230.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700020706.mm.1 TC0700002308.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 514302 - -
          ABI Mouse Genome Survey Microarray 757892 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3840070 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840070 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2443381 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2443381 - -
          mouse-6 v1.1 (Illumina) scl31762.1.248_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840070 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31762.1.248_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840070 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2443381 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2443381 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline