| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500025196.mm.1 |
NM_144955.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025196.mm.1 |
NM_144955.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025196.mm.1 |
TC0500002856.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025195.mm.1 |
NM_144955.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025195.mm.1 |
NM_144955.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025195.mm.1 |
TC0500002856.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500025197.mm.1 |
NM_144955.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025197.mm.1 |
NM_144955.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500025197.mm.1 |
TC0500002856.mm.1 |
MTA TC |