Search
Search

Oasl1| Gene Symbol: Oasl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145209.3 | NP_660210.1 | 3 - 4 | 577 | 72 | |
| GenBank mRNA | AB067533.1 | - | 2 - 3 | 502 | 72 | |
| AF426289.1 | - | 2 - 3 | 508 | 72 | ||
| AK078690.1 | - | 3 - 4 | 578 | 72 | ||
| AK089111.1 | - | 2 - 3 | 547 | 72 | ||
| AK149824.1 | - | 2 - 3 | 501 | 72 | ||
| AK150863.1 | - | 2 - 3 | 631 | 72 | ||
| AK165578.1 | - | 2 - 3 | 614 | 72 | ||
| AY057107.1 | - | 2 - 3 | 490 | 72 | ||
| AY089728.1 | - | 2 - 3 | 502 | 72 | ||
| BC032152.1 | - | 2 - 3 | 514 | 5068699 | 72 | |
Set Membership: |
|