Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455104_m1
          See other RIN1 GE Assays ›
          Gene Symbol:
          Rin1
          Gene Name:
          Ras and Rab interactor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 5050010 - 5057072 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455104_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145495.2 NP_663470.1 9 - 10 1889 105
          GenBank mRNA BC011277.1 - 9 - 10 1889 3711198 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000054.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rin1

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          endocytosis
          signal transduction
          memory
          associative learning
          negative regulation of synaptic plasticity
          GTPase activator activity
          Ras GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211765 NM_145495.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211765 BC011277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211765 NM_145495.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P211765 BC011277.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292972 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735990 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148059 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211765 NM_145495.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211765 BC011277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211765 NM_145495.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P211765 BC011277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211765 NM_145495.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211765 BC011277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211765 NM_145495.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P211765 BC011277.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31926585 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03542312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148059 - -
          Whole Mouse Genome G4122A A_51_P211765 NM_145495.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211765 BC011277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211765 NM_145495.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P211765 BC011277.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148059 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14209570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14209552 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14209559 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14209581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14209580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14209548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31926499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14504363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14504359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14504358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31926598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14209534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14177361 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460538 - -
          Affymetrix HT MG-430 PM Array Plate 1437460_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1424507_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460529 - -
          Mouse Genome 430 2.0 Array 1424507_at - -
          Mouse Genome 430 2.0 Array 1437460_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437460_x_at - -
          Mouse Genome 430 2.0 Array 1424507_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225870_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225870_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900000725.mm.1 NM_145495.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000725.mm.1 NM_145495.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000725.mm.1 TC1900000054.mm.1 MTA TC
          Mouse Expression Set 430 1424507_PM_at - -
          Mouse Expression Set 430 1424507_at - -
          Mouse Expression Set 430 1437460_x_at - -
          Mouse Expression Set 430 1437460_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1424507_PM_at - -
          Mouse Genome 430A 2.0 Array 1424507_at - -
          Affymetrix HT Mouse Genome 430A Array 1424507_at - -
          Affymetrix HT Mouse Genome 430A Array 1424507_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437460_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1437460_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 478326 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102760450 - -
          Illumina mouse-6 v1.0 expression beadchip 102760450 - -
          Illumina mouse-6 v1.0 expression beadchip 510593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2650289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2650289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260395 - -
          mouse-6 v1.1 (Illumina) scl0225870.1_275-S - -
          mouse-6 v1.1 (Illumina) GI_21703973-S - -
          Illumina Sentrix mouse-6.v1 bead array 510593 - -
          Illumina Sentrix mouse-6.v1 bead array 102760450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0225870.1_275-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102760450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2650289 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2650289 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0225870.1_275-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline