Search
Search
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145614.4 | NP_663589.3 | 7 - 8 | 1429 | 71 | |
| GenBank mRNA | AK032124.1 | - | 7 - 8 | 1430 | 71 | |
| AK043771.1 | - | 7 - 8 | 1118 | 71 | ||
| AK166090.1 | - | 7 - 8 | 1199 | 71 | ||
| AY044265.1 | - | 7 - 8 | 866 | 71 | ||
| BC026680.1 | - | 7 - 8 | 1177 | 5059786 | 71 | |
| BC031495.1 | - | 7 - 8 | 1150 | 2651095 | 71 | |
| BC069862.1 | - | 7 - 8 | 1172 | 5388446 | 71 | |
Set Membership: |
|