Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455226_m1
          See other ADIG GE Assays ›
          Gene Symbol:
          Adig
          Gene Name:
          adipogenin
          Gene Aliases:
          SMAF-1, Smaf1
          Chromosome Location:
          Chr.2: 158502612 - 158508198 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455226_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adig
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145635.2 NP_663610.1 2 - 3 383 98
          GenBank mRNA AK131610.1 - 2 - 3 333 98
          AY092026.1 - 2 - 3 389 98
          BC054059.1 - 2 - 3 331 3373967 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002435.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adig

          Gene Ontology Categories:

          Function(s) Process(es)

          fat cell differentiation
          positive regulation of fat cell differentiation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182716 NM_145635.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182716 AK131610.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182716 AY092026.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182716 BC054059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2182716 BE624674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042500 NM_145635.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642808 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134208 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182716 NM_145635.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182716 AK131610.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182716 AY092026.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182716 BC054059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2182716 BE624674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042500 NM_145635.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21783275 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00539028 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134208 - -
          Whole Mouse Genome G4122A A_51_P505738 AY092026.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137340 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01803681 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01803673 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01803680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01803676 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01448497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01803682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21783352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02198920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01944214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01623528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01803676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01917437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02176456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01497627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01416455 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02149530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01448499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01448497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21783273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478113 - -
          Affymetrix HT MG-430 PM Array Plate 1424729_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478109 - -
          Mouse Genome 430 2.0 Array 1424729_at - -
          Mouse Genome 430 2.0 Array 1424729_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246747_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 246747_at - -
          Affymetrix Mouse Clariom S Array PSR0200019789.mm.1 NM_145635.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200019789.mm.1 NM_145635.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200019789.mm.1 TC0200002435.mm.1 MTA TC
          Mouse Expression Set 430 1424729_PM_at - -
          Mouse Expression Set 430 1424729_at - -
          Murine Genome U74v2 Set 96237_PM_at - -
          Murine Genome U74v2 Set 96237_at - -
          Mouse Genome 430A 2.0 Array 1424729_PM_at - -
          Mouse Genome 430A 2.0 Array 1424729_at - -
          Affymetrix HT Mouse Genome 430A Array 1424729_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424729_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 323222 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780672 - -
          Illumina mouse-6 v1.0 expression beadchip 780672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692644 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692644 - -
          mouse-6 v1.1 (Illumina) scl19994.5.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 780672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19994.5.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692644 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline