Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455277_s1
          See other VMN1R85 GE Assays ›
          Gene Symbol:
          Vmn1r85
          Gene Name:
          vomeronasal 1 receptor 85
          Gene Aliases:
          V1rj3
          Chromosome Location:
          Chr.7: 13084289 - 13085215 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455277_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145847.1 NP_665846.1 1 648 73
          GenBank mRNA BC110463.1 - 1 648 40040729 73
          BC110464.1 - 1 648 40040731 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002362.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r85

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226527 NM_145847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226527 BC110463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226527 BC110464.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226527 NM_145847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226527 BC110463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226527 BC110464.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157360 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226527 NM_145847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226527 BC110463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226527 BC110464.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226527 NM_145847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226527 BC110463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226527 BC110464.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226527 NM_145847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226527 BC110463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226527 BC110464.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226527 NM_145847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226527 BC110463.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226527 BC110464.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24974524 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157360 - -
          Whole Mouse Genome G4122A A_51_P226527 NM_145847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226527 BC110463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226527 BC110464.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226527 NM_145847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226527 BC110463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226527 BC110464.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114400 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24974497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06311947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06345963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06575324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06345962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06311930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06184017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06168039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06345965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06345959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06101281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06500702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06345956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24974518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560088 - -
          Affymetrix Mouse Gene 1.1 ST Array 10560087 - -
          Affymetrix Mouse Clariom S Array PSR0700020994.mm.1 TC0700002362.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700020994.mm.1 NM_145847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700020994.mm.1 NM_145847.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 446321 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870204 - -
          Illumina mouse-6 v1.0 expression beadchip 870204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2417190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2417190 - -
          mouse-6 v1.1 (Illumina) scl9197.1.1_237-S - -
          Illumina Sentrix mouse-6.v1 bead array 870204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9197.1.1_237-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2417190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2417190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline