Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455528_s1
          See other OLFR25 GE Assays ›
          Gene Symbol:
          Olfr25
          Gene Name:
          olfactory receptor 25
          Gene Aliases:
          MOR170-4, MTPCR18
          Chromosome Location:
          Chr.9: 38329492 - 38330559 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455528_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146870.2 NP_667081.2 1 571 75
          GenBank mRNA BC125306.1 - 1 571 40129821 75
          BC125308.1 - 1 571 40129823 75
          X89684.1 - 1 91 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000394.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr25

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078790 BC125306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078790 BC125308.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2078790 NM_146870.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078790 BC125306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078790 BC125308.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2078790 NM_146870.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225815 BC125306.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225815 BC125308.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225815 NM_146870.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P225815 X89684.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434363 - -
          Whole Mouse Genome G4122A A_51_P225815 BC125306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225815 BC125308.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225815 NM_146870.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225815 X89684.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140248 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07748278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07748272 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07748287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07748286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08020547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08101302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07748287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07901360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584398 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584395 - -
          Affymetrix Mouse Clariom S Array PSR0900002790.mm.1 NM_146870.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002790.mm.1 NM_146870.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002790.mm.1 TC0900000394.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 372981 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5270022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239971 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239971 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0