Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455619_s1
          See other OLFR653 GE Assays ›
          Gene Symbol:
          Olfr653
          Gene Name:
          olfactory receptor 653
          Gene Aliases:
          MOR33-1, Ors19
          Chromosome Location:
          Chr.7: 104579624 - 104580669 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455619_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr653
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147074.2 NP_667285.2 1 71 108
          GenBank mRNA BC116962.1 - 1 71 40126281 108
          BC120700.1 - 1 71 40129623 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001525.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr653

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P172048 BC120700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P172048 NM_147074.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P172048 BC116962.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P172048 BC120700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P172048 NM_147074.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P172048 BC116962.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263776 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171875 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154250 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135714 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P172048 BC120700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P172048 NM_147074.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P172048 BC116962.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P172048 BC120700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P172048 NM_147074.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P172048 BC116962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172048 BC120700.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172048 NM_147074.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172048 BC116962.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172048 BC120700.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172048 NM_147074.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P172048 BC116962.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135714 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154250 - -
          Whole Mouse Genome G4122A A_51_P172048 BC120700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172048 NM_147074.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172048 BC116962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172048 BC120700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172048 NM_147074.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P172048 BC116962.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139582 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135714 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154250 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25388688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05924833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05924827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05966217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25388629 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555876 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555875 - -
          Affymetrix Mouse Clariom S Array PSR0700012894.mm.1 TC0700001525.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012894.mm.1 NM_147074.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012894.mm.1 NM_147074.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379908 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4780465 - -
          Illumina mouse-6 v1.0 expression beadchip 4780465 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780465 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745912 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745912 - -
          mouse-6 v1.1 (Illumina) scl9386.1.1_133-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780465 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9386.1.1_133-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780465 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745912 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745912 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline