Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455863_m1
          See other UBQLN1 GE Assays ›
          Gene Symbol:
          Ubqln1
          Gene Name:
          ubiquilin 1
          Gene Aliases:
          1110046H03Rik, 1810030E05Rik, AU019746, C77538, D13Ertd372e, Da41, Dsk2, Plic-1, Plic1, Xdrp1
          Chromosome Location:
          Chr.13: 58176156 - 58215653 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455863_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ubqln1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026842.4 NP_081118.4 2 - 3 526 70
          NM_152234.2 NP_689420.1 2 - 3 526 70
          GenBank mRNA AK048534.1 - 2 - 3 447 70
          AK049298.1 - 2 - 3 281 70
          AK165456.1 - 2 - 3 491 70
          AK172695.1 - 2 - 3 495 70
          BC026847.1 - 2 - 3 509 4015641 70
          BC028857.1 - 2 - 3 465 4507759 70
          BC051098.1 - 1 - 2 154 70
          BC080667.1 - 2 - 3 487 4935292 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002095.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ubqln1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of macroautophagy
          ER-associated ubiquitin-dependent protein catabolic process
          regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          response to endoplasmic reticulum stress
          aggrephagy
          cellular response to hypoxia
          negative regulation of autophagosome maturation
          negative regulation of store-operated calcium channel activity
          regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          positive regulation of ER-associated ubiquitin-dependent protein catabolic process
          regulation of autophagosome assembly
          receptor binding
          intermediate filament binding
          kinase binding
          protein domain specific binding
          polyubiquitin binding
          identical protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300013309.mm.1 NM_026842.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013309.mm.1 NM_026842.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013309.mm.1 NM_152234.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013309.mm.1 NM_152234.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013309.mm.1 TC1300002095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013311.mm.1 NM_026842.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013311.mm.1 NM_026842.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013311.mm.1 NM_152234.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013311.mm.1 NM_152234.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013311.mm.1 TC1300002095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013312.mm.1 NM_026842.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013312.mm.1 NM_026842.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013312.mm.1 NM_152234.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013312.mm.1 NM_152234.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013312.mm.1 TC1300002095.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013310.mm.1 NM_026842.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013310.mm.1 NM_026842.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013310.mm.1 NM_152234.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013310.mm.1 NM_152234.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013310.mm.1 TC1300002095.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931003 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline