Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00456011_m1
          See other CTSG GE Assays ›
          Gene Symbol:
          Ctsg
          Gene Name:
          cathepsin G
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 56099881 - 56102574 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00456011_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctsg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007800.1 NP_031826.1 1 - 2 187 70
          GenBank mRNA BC125511.1 - 1 - 2 102 40130026 70
          BC125513.1 - 1 - 2 102 40130028 70
          X78544.1 - 1 - 2 187 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002115.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protease Mammalian Gene Collection Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctsg

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          proteolysis
          immune response
          protein processing
          response to lipopolysaccharide
          negative regulation of growth of symbiont in host
          positive regulation of immune response
          defense response to Gram-positive bacterium
          defense response to fungus
          neutrophil mediated killing of gram-positive bacterium
          serine-type endopeptidase activity
          heparin binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 BC125513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 X78544.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 BC125511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 NM_007800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 BC125513.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 X78544.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 BC125511.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P416546 NM_007800.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1844 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170340 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 BC125513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 X78544.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 BC125511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 NM_007800.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 BC125513.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 X78544.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 BC125511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P416546 NM_007800.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 BC125513.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 X78544.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 BC125511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 NM_007800.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 BC125513.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 X78544.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 BC125511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P416546 NM_007800.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29621442 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170340 - -
          Whole Mouse Genome G4122A A_51_P416546 BC125513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 X78544.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 BC125511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 NM_007800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 BC125513.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 X78544.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 BC125511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P416546 NM_007800.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136416 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106500 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170340 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657431 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657425 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657424 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11657434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29621417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11655533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621434 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11657425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11657424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11655530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11764104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11524890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11165381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11657435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621403 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11657429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11764101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29621454 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10420263 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420266 - -
          Affymetrix Mouse Gene 1.0 ST Array 10420262 - -
          Affymetrix HT MG-430 PM Array Plate 1419594_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10420261 - -
          Mouse Genome 430 2.0 Array 1419594_PM_at - -
          Mouse Genome 430 2.0 Array 1419594_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13035_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13035_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400016291.mm.1 NM_007800.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016291.mm.1 TC1400002115.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016290.mm.1 NM_007800.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016290.mm.1 TC1400002115.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400016289.mm.1 NM_007800.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400016289.mm.1 TC1400002115.mm.1 MTA TC
          Mouse Expression Set 430 1419594_at - -
          Mouse Expression Set 430 1419594_PM_at - -
          Murine Genome U74v2 Set 92924_at - -
          Murine Genome U74v2 Set 92924_PM_at - -
          Mouse Genome 430A 2.0 Array 1419594_at - -
          Mouse Genome 430A 2.0 Array 1419594_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419594_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419594_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545933 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220236 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220236 - -
          mouse-6 v1.1 (Illumina) scl45515.5.1_34-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45515.5.1_34-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220236 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220236 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline