Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00456405_m1
          See other ZFP57 GE Assays ›
          Gene Symbol:
          Zfp57
          Gene Name:
          zinc finger protein 57
          Gene Aliases:
          G19, Zfp-57
          Chromosome Location:
          Chr.17: 37001163 - 37010729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00456405_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp57
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013745.2 NP_001013767.1 4 - 5 531 77
          NM_001168501.1 NP_001161973.1 4 - 5 397 77
          NM_001168502.1 NP_001161974.1 5 - 6 773 77
          NR_033137.1 2 - 3 1379 77
          GenBank mRNA AK054083.1 - 4 - 5 514 77
          AK135881.1 - 4 - 5 504 77
          AK136241.1 - 4 - 5 515 77
          AK139625.1 - 5 - 6 773 77
          AK143118.1 - 2 - 3 1379 77
          AK145368.1 - 4 - 5 503 77
          BC052028.1 - 2 - 3 698 5686931 77
          D21850.1 - 3 - 4 526 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000752.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp57

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          DNA methylation involved in embryo development
          nucleic acid binding
          DNA binding
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029533 BC052028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029533 NR_033137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029533 NM_001013745.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029533 NM_001168501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2029533 NM_001168502.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172201 AK145368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172201 BC052028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172201 D21850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172201 AK143118.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172201 AK136241.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 90717 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103886 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029533 BC052028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029533 NR_033137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029533 NM_001013745.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029533 NM_001168501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2029533 NM_001168502.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172201 AK145368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172201 BC052028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172201 D21850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172201 AK143118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172201 AK136241.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197925 AK145368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197925 BC052028.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197925 D21850.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197925 AK143118.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P197925 AK136241.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166713 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08772018 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103886 - -
          Whole Mouse Genome G4122A A_51_P197925 AK145368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197925 BC052028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197925 D21850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197925 AK143118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P197925 AK136241.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13196932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13196935 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13196936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13172527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13460300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13255602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13469854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13172531 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445075 - -
          Affymetrix HT MG-430 PM Array Plate 1450929_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445071 - -
          Mouse Genome 430 2.0 Array 1450929_PM_at - -
          Mouse Genome 430 2.0 Array 1450929_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22715_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22715_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NM_001013745.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NM_001013745.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NM_001168501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NM_001168501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NM_001168502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NM_001168502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NR_033137.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 NR_033137.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007610.mm.1 TC1700000752.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NM_001013745.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NM_001013745.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NM_001168501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NM_001168501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NM_001168502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NM_001168502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NR_033137.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 NR_033137.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007613.mm.1 TC1700000752.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NM_001013745.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NM_001013745.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NM_001168501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NM_001168501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NM_001168502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NM_001168502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NR_033137.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 NR_033137.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007607.mm.1 TC1700000752.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NM_001013745.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NM_001013745.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NM_001168501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NM_001168501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NM_001168502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NM_001168502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NR_033137.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 NR_033137.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007609.mm.1 TC1700000752.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NM_001013745.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NM_001013745.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NM_001168501.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NM_001168501.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NM_001168502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NM_001168502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NR_033137.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 NR_033137.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007608.mm.1 TC1700000752.mm.1 MTA TC
          Mouse Expression Set 430 1450929_PM_at - -
          Mouse Expression Set 430 1450929_at - -
          Murine Genome U74v2 Set 103437_PM_at - -
          Murine Genome U74v2 Set 103437_at - -
          Mouse Genome 430A 2.0 Array 1450929_PM_at - -
          Mouse Genome 430A 2.0 Array 1450929_at - -
          Affymetrix HT Mouse Genome 430A Array 1450929_at - -
          Affymetrix HT Mouse Genome 430A Array 1450929_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 921996 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221161 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221161 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221161 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221161 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline