Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00456954_s1
          See other ZNRF4 GE Assays ›
          Gene Symbol:
          Znrf4
          Gene Name:
          zinc and ring finger 4
          Gene Aliases:
          1700094M01Rik, Ssrzf1, spzn
          Chromosome Location:
          Chr.17: 56511248 - 56512483 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00456954_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Znrf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011483.2 NP_035613.2 1 720 90
          GenBank mRNA AB016984.1 - 1 654 90
          AK005843.1 - 1 669 90
          BC140986.1 - 1 603 9055598 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002328.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Znrf4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256675 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256676 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256665 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256673 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256671 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256666 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256677 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977692 AB016984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977692 AK005843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977692 NM_011483.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977692 BC140986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977692 AB016984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977692 AK005843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977692 NM_011483.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977692 BC140986.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266977 AB016984.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31256716 - -
          Whole Mouse Genome G4122A A_51_P266977 AB016984.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13065045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13065038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13065044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13065040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13065043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13065034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256677 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256671 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256665 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256675 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256673 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256666 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31256676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31256680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13065032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13407220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13476206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13065048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13217095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13193193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13361867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13227271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13392090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13579624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13538428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13065027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256693 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13476208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13227267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13538429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31256642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13343843 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10452084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452083 - -
          Affymetrix HT MG-430 PM Array Plate 1449535_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10452082 - -
          Mouse Genome 430 2.0 Array 1449535_PM_at - -
          Mouse Genome 430 2.0 Array 1449535_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20834_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20834_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700021763.mm.1 NM_011483.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021763.mm.1 NM_011483.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021763.mm.1 TC1700002328.mm.1 MTA TC
          Mouse Expression Set 430 1449535_at - -
          Mouse Expression Set 430 1449535_PM_at - -
          Murine Genome U74v2 Set 98310_at - -
          Murine Genome U74v2 Set 98310_PM_at - -
          Mouse Genome 430A 2.0 Array 1449535_at - -
          Mouse Genome 430A 2.0 Array 1449535_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449535_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449535_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 665024 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5690750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225919 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225919 - -
          mouse-6 v1.1 (Illumina) scl49764.1.12_287-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49764.1.12_287-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225919 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225919 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline