Search
Search
| Gene Symbol: Orc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177313.1 | NP_001170784.1 | 2 - 3 | 224 | 82 | |
| NM_011958.3 | NP_036088.3 | 2 - 3 | 224 | 82 | ||
| NR_033442.1 | 2 - 3 | 224 | 82 | |||
| GenBank mRNA | AJ003140.1 | - | 2 - 3 | 185 | 82 | |
| AK010258.1 | - | 2 - 3 | 208 | 82 | ||
| AK011550.1 | - | 2 - 3 | 190 | 82 | ||
| AK012669.1 | - | 2 - 3 | 192 | 82 | ||
| AK049534.1 | - | 2 - 3 | 192 | 82 | ||
| AK076324.1 | - | 2 - 3 | 225 | 82 | ||
| AK084326.1 | - | 2 - 3 | 193 | 82 | ||
| AK145454.1 | - | 2 - 3 | 235 | 82 | ||
| AK169291.1 | - | 2 - 3 | 208 | 82 | ||
| BC015072.1 | - | 2 - 3 | 196 | 4212266 | 82 | |
| BC019748.1 | - | 2 - 3 | 166 | 5007346 | 82 | |
| Y16386.1 | - | 1 - 2 | 56 | 82 | ||
Set Membership: |
|