Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00457412_m1
          See other PRSS30 GE Assays ›
          Gene Symbol:
          Prss30
          Gene Name:
          protease, serine 30
          Gene Aliases:
          2010015P21Rik, Disp, Disp1, Tmprss8
          Chromosome Location:
          Chr.17: 23972127 - 23980572 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00457412_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss30
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013921.3 NP_038949.2 1 - 2 103 66
          GenBank mRNA AJ243866.1 - 1 - 2 66 66
          BC010970.1 - 1 - 2 96 4216644 66
          BC040348.1 - 1 - 2 103 4206523 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss30

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          transport
          ion transport
          sodium ion transport
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          sodium channel regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P241935 AJ243866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P241935 AJ243866.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294031 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173784 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P241935 AJ243866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P241935 AJ243866.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241935 AJ243866.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P241935 AJ243866.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31094656 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173784 - -
          Whole Mouse Genome G4122A A_51_P241935 AJ243866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P241935 AJ243866.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123031 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13097755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13097740 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13097759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13097746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13097751 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13097738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13083715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13571173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13460289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13375197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13381073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13535243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13078958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13097741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13083717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13548032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13195428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13097751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13523724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13305505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13548029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13302203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13548031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13097740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13097755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094662 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448403 - -
          Affymetrix HT MG-430 PM Array Plate 1449952_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448402 - -
          Mouse Genome 430 2.0 Array 1449952_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449952_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30943_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30943_PM_at - -
          Mouse Expression Set 430 1449952_s_at - -
          Mouse Expression Set 430 1449952_PM_s_at - -
          Murine Genome U74v2 Set 93709_PM_at - -
          Murine Genome U74v2 Set 93709_at - -
          Mouse Genome 430A 2.0 Array 1449952_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449952_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449952_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449952_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463972 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6620575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006620575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258386 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258386 - -
          mouse-6 v1.1 (Illumina) scl50223.6.1_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 6620575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50223.6.1_69-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258386 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline