Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00457645_m1
          See other IL1F6 GE Assays ›
          Gene Symbol:
          Il1f6
          Gene Name:
          interleukin 1 family, member 6
          Gene Aliases:
          Fil1, IL-1H1, IL1RP2, Il1f9, Il36a
          Chromosome Location:
          Chr.2: 24215417 - 24225701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00457645_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1f6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019450.3 NP_062323.1 4 - 5 430 103
          GenBank mRNA AF200493.1 - 3 - 4 269 103
          AF206697.1 - 4 - 5 344 103
          AK004061.1 - 4 - 5 430 103
          AK224931.1 - 4 - 5 364 103
          AK224932.1 - 4 - 5 377 103
          BC117102.1 - 3 - 4 303 40126421 103
          BC117104.1 - 3 - 4 303 40126423 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000377.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il1f6

          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          immune system process
          inflammatory response
          immune response
          cytokine-mediated signaling pathway
          positive regulation of interleukin-6 production
          innate immune response
          cytokine activity
          interleukin-1 receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109708 AK224928.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109708 AK224929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109708 AK224931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109708 NM_019450.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P109708 AF206697.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 309065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118260 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135457 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171185 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109708 AK224928.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109708 AK224929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109708 AK224931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109708 NM_019450.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109708 AF206697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504194 AK224931.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504194 AK224932.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504194 BC117102.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504194 BC117104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504194 NM_019450.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171185 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135457 - -
          Whole Mouse Genome G4122A A_51_P504194 AK224931.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504194 AK224932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504194 BC117102.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504194 BC117104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504194 NM_019450.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109708 AK224928.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109708 AK224929.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109708 AK224931.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109708 NM_019450.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109708 AF206697.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119577 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171185 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135457 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01207441 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02272133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02272134 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02272144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01634850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02272140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02272152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064661 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469794 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469796 - -
          Affymetrix HT MG-430 PM Array Plate 1418609_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469793 - -
          Mouse Genome 430 2.0 Array 1418609_at - -
          Mouse Genome 430 2.0 Array 1418609_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54448_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 54448_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200001881.mm.1 NM_019450.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200001881.mm.1 NM_019450.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200001881.mm.1 TC0200000377.mm.1 MTA TC
          Mouse Expression Set 430 1418609_PM_at - -
          Mouse Expression Set 430 1418609_at - -
          Mouse Genome 430A 2.0 Array 1418609_at - -
          Mouse Genome 430A 2.0 Array 1418609_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418609_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418609_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 668748 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3450138 - -
          Illumina mouse-6 v1.0 expression beadchip 3450138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1251134 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1251134 - -
          mouse-6 v1.1 (Illumina) scl21203.6.1_140-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21203.6.1_140-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1251134 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251134 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline