Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00457884_s1
          See other ZBTB22 GE Assays ›
          Gene Symbol:
          Zbtb22
          Gene Name:
          zinc finger and BTB domain containing 22
          Gene Aliases:
          1110008J20Rik, AI265210, AI415166, Bing1, Zfp297
          Chromosome Location:
          Chr.17: 33915895 - 33919325 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00457884_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020625.3 NP_065650.1 2 318 69
          GenBank mRNA AK155199.1 - 2 302 69
          AK157161.1 - 2 309 69
          AK159082.1 - 2 308 69
          AK159789.1 - 2 309 69
          BC026964.1 - 2 296 5043642 69
          BC052154.1 - 2 265 6329405 69
          BY011334.1 - 2 319 69
          KJ650209.1 - 1 154 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002777.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb22

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150957 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150956 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 BC052154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 AK159789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 AK202169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 BC026964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 AK157161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 BC052154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 AK159789.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 AK202169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 BC026964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P365005 AK157161.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304631 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880644 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146970 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 BC052154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 AK159789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 AK202169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 BC026964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 AK157161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 BC052154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 AK159789.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 AK202169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 BC026964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P365005 AK157161.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31150977 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146970 - -
          Whole Mouse Genome G4122A A_52_P365005 BC052154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 AK159789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 AK202169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 BC026964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 AK157161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 BC052154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 AK159789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 AK202169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 BC026964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P365005 AK157161.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126853 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146970 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450832 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125651 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125647 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13125656 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450825 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13463798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450831 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13450785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150957 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31150955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31150980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13450815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13450795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31150973 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444067 - -
          Affymetrix HT MG-430 PM Array Plate 1448705_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444066 - -
          Mouse Genome 430 2.0 Array 1448705_at - -
          Mouse Genome 430 2.0 Array 1448705_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81630_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81630_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700005542.mm.1 NM_020625.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005542.mm.1 NM_020625.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005542.mm.1 TC1700002777.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005543.mm.1 NM_020625.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005543.mm.1 NM_020625.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005543.mm.1 TC1700002777.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005540.mm.1 NM_020625.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005540.mm.1 NM_020625.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005540.mm.1 TC1700002777.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700005541.mm.1 NM_020625.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005541.mm.1 NM_020625.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700005541.mm.1 TC1700002777.mm.1 MTA TC
          Mouse Expression Set 430 1448705_at - -
          Mouse Expression Set 430 1448705_PM_at - -
          Murine Genome U74v2 Set 100897_PM_f_at - -
          Murine Genome U74v2 Set 100897_f_at - -
          Mouse Genome 430A 2.0 Array 1448705_PM_at - -
          Mouse Genome 430A 2.0 Array 1448705_at - -
          Affymetrix HT Mouse Genome 430A Array 1448705_at - -
          Affymetrix HT Mouse Genome 430A Array 1448705_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 396014 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 580463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000580463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233129 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233129 - -
          mouse-6 v1.1 (Illumina) scl081630.1_308-S - -
          Illumina Sentrix mouse-6.v1 bead array 580463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl081630.1_308-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 580463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233129 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233129 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl081630.1_308-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline