Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00458148_m1
          See other ZFP296 GE Assays ›
          Gene Symbol:
          Zfp296
          Gene Name:
          zinc finger protein 296
          Gene Aliases:
          2210018A16Rik
          Chromosome Location:
          Chr.7: 19577287 - 19580656 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00458148_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp296
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022409.2 NP_071854.2 2 - 3 484 66
          GenBank mRNA AJ278340.1 - 1 - 2 143 66
          BC099454.1 - 2 - 3 410 30936046 66
          BC107243.2 - 1 - 2 130 66
          BE635089.1 - 1 - 2 61 66
          DV647330.1 - 2 - 3 484 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000305.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp296

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor zinc finger transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011026 AK008748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011026 BE634620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011026 AJ278340.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011026 NM_022409.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011026 AF450095.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482061 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180306 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157451 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011026 AK008748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011026 BE634620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011026 AJ278340.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011026 NM_022409.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011026 AF450095.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24994134 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180306 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157451 - -
          Whole Mouse Genome G4122A A_51_P154987 BC099454.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154987 BC107243.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154987 BE634620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154987 AJ278340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154987 NM_022409.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154983 AK008748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154983 BC099454.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154983 BE634620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154983 AJ278340.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154983 NM_022409.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123896 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157451 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180306 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06110064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24994121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06385652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06110066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06110075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06110081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06385650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24994142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978113 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550733 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550732 - -
          Affymetrix HT MG-430 PM Array Plate 1449231_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550730 - -
          Mouse Genome 430 2.0 Array 1449231_PM_at - -
          Mouse Genome 430 2.0 Array 1449231_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 63872_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 63872_at - -
          Affymetrix Mouse Clariom S Array PSR0700002445.mm.1 TC0700000305.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002445.mm.1 NM_022409.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002445.mm.1 NM_022409.2 Refseq
          Mouse Expression Set 430 1449231_PM_at - -
          Mouse Expression Set 430 1449231_at - -
          Murine Genome U74v2 Set 170935_PM_r_at - -
          Murine Genome U74v2 Set 170935_r_at - -
          Mouse Genome 430A 2.0 Array 1449231_PM_at - -
          Mouse Genome 430A 2.0 Array 1449231_at - -
          Affymetrix HT Mouse Genome 430A Array 1449231_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449231_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 435181 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5080180 - -
          Illumina mouse-6 v1.0 expression beadchip 5080180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005080180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2470407 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2470407 - -
          mouse-6 v1.1 (Illumina) scl32990.3.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 5080180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32990.3.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5080180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2912729 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2470407 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2912729 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2470407 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32990.3.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline