Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00458451_m1
          See other FOXI1 GE Assays ›
          Gene Symbol:
          Foxi1
          Gene Name:
          forkhead box I1
          Gene Aliases:
          5830401E05Rik, FREAC6, Fkh10, HFH-3, Hfh3
          Chromosome Location:
          Chr.11: 34204341 - 34208089 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00458451_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxi1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023907.4 NP_076396.3 1 - 2 622 61
          GenBank mRNA AK019994.1 - 1 - 2 624 61
          BC007475.1 - 1 - 2 587 3601768 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002496.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Foxi1

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          inner ear morphogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166513 AK019994.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166513 BC007475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166513 NM_023907.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965582 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128347 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166513 AK019994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166513 BC007475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166513 NM_023907.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391207 AK019994.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391207 BC007475.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P391207 NM_023907.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27699561 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128347 - -
          Whole Mouse Genome G4122A A_51_P391207 AK019994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391207 BC007475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391207 NM_023907.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120257 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09558456 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09558462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09558458 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09558461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09636462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27699546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08997157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08997177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09728414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09447043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09316778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09558456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09612926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09636462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09316785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27699606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385115 - -
          Affymetrix HT MG-430 PM Array Plate 1449458_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385114 - -
          Mouse Genome 430 2.0 Array 1449458_PM_at - -
          Mouse Genome 430 2.0 Array 1449458_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14233_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14233_at - -
          Affymetrix Mouse Clariom S Array PSR1100023260.mm.1 NM_023907.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023260.mm.1 NM_023907.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023260.mm.1 TC1100002496.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023262.mm.1 NM_023907.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023262.mm.1 NM_023907.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023262.mm.1 TC1100002496.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023261.mm.1 NM_023907.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023261.mm.1 NM_023907.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023261.mm.1 TC1100002496.mm.1 MTA TC
          Mouse Expression Set 430 1449458_at - -
          Mouse Expression Set 430 1449458_PM_at - -
          Murine Genome U74v2 Set 135167_PM_at - -
          Murine Genome U74v2 Set 135167_at - -
          Mouse Genome 430A 2.0 Array 1449458_PM_at - -
          Mouse Genome 430A 2.0 Array 1449458_at - -
          Affymetrix HT Mouse Genome 430A Array 1449458_at - -
          Affymetrix HT Mouse Genome 430A Array 1449458_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 866808 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6620195 - -
          Illumina mouse-6 v1.0 expression beadchip 3710095 - -
          Illumina mouse-6 v1.0 expression beadchip 6620195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006620195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722233 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658723 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658723 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722233 - -
          mouse-6 v1.1 (Illumina) scl0001286.1_73-S - -
          mouse-6 v1.1 (Illumina) scl40370.2_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710095 - -
          Illumina Sentrix mouse-6.v1 bead array 6620195 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40370.2_46-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001286.1_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710095 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620195 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658723 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3000416 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722232 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722232 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658723 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3000416 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline