Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00458558_s1
          See other ACTL7B GE Assays ›
          Gene Symbol:
          Actl7b
          Gene Name:
          actin-like 7b
          Gene Aliases:
          Tact1
          Chromosome Location:
          Chr.4: 56740005 - 56741425 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00458558_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actl7b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025271.2 NP_079547.2 1 1267 57
          GenBank mRNA AB023062.1 - 1 1225 57
          AF113520.1 - 1 1250 57
          BC116815.1 - 1 1219 40126134 57
          BC116817.1 - 1 1219 40126136 57
          BX634818.1 - 1 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002704.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Actl7b

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22941198 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22941196 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22941195 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 BC116815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 BC116817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 NM_025271.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 AB023062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 AF113520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 BC116815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 BC116817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 NM_025271.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 AB023062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471619 AF113520.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 761084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954584 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117841 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 BC116815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 BC116817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 NM_025271.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 AB023062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 AF113520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 BC116815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 BC116817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 NM_025271.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 AB023062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471619 AF113520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 BC116815.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 BC116817.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 NM_025271.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 AB023062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 AF113520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 BC116815.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 BC116817.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 NM_025271.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 AB023062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P471619 AF113520.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22941199 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117841 - -
          Whole Mouse Genome G4122A A_51_P471619 BC116815.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 BC116817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 NM_025271.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 AB023062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 AF113520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 BC116815.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 BC116817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 NM_025271.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 AB023062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471619 AF113520.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101586 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03830879 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03845117 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03473975 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03473971 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04007050 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03473956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03473981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22941196 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22941198 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22941195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03473950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941195 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03829093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10513018 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513017 - -
          Affymetrix HT MG-430 PM Array Plate 1424678_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10513016 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505105 - -
          Mouse Genome 430 2.0 Array 1424678_PM_at - -
          Mouse Genome 430 2.0 Array 1424678_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11471_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11471_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400021776.mm.1 TC0400002704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021776.mm.1 NM_025271.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021776.mm.1 NM_025271.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021775.mm.1 TC0400002704.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021775.mm.1 NM_025271.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021775.mm.1 NM_025271.2 Refseq
          Mouse Expression Set 430 1424678_at - -
          Mouse Expression Set 430 1424678_PM_at - -
          Murine Genome U74v2 Set 101864_PM_at - -
          Murine Genome U74v2 Set 101864_at - -
          Mouse Genome 430A 2.0 Array 1424678_at - -
          Mouse Genome 430A 2.0 Array 1424678_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424678_at - -
          Affymetrix HT Mouse Genome 430A Array 1424678_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 790039 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100070687 - -
          MouseWG-6 v2 Expression BeadChip 460341 - -
          Illumina mouse-6 v1.0 expression beadchip 460341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2675497 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2675497 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2547689 - -
          mouse-6 v1.1 (Illumina) scl24311.1.14_313-S - -
          Illumina Sentrix mouse-6.v1 bead array 460341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24311.1.14_313-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2675497 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2675497 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline