Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00458573_m1
          See other AKR7A5 GE Assays ›
          Gene Symbol:
          Akr7a5
          Gene Name:
          aldo-keto reductase family 7, member A5 (aflatoxin aldehyde reductase)
          Gene Aliases:
          0610025K21Rik, Afar, Afar1, Akr7a2
          Chromosome Location:
          Chr.4: 139310738 - 139318786 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00458573_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr7a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025337.3 NP_079613.3 3 - 4 630 101
          GenBank mRNA AF525358.2 - 3 - 4 532 101
          AJ271800.1 - 3 - 4 547 101
          AJ271801.1 - 3 - 4 547 101
          AK002664.1 - 3 - 4 610 101
          AK003915.1 - 3 - 4 617 101
          AK143203.1 - 3 - 4 629 101
          BC031857.1 - 3 - 4 607 5124841 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001743.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr7a5

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          daunorubicin metabolic process
          doxorubicin metabolic process
          oxidation-reduction process
          alditol:NADP+ 1-oxidoreductase activity
          oxidoreductase activity
          phenanthrene-9,10-epoxide hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351040 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351041 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 AK002664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 AK003915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 AK143203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 BC031857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 NM_025337.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 AK002664.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 AK003915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 AK143203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 BC031857.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P188795 NM_025337.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 770851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 617504 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164675 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 AK002664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 AK003915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 AK143203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 BC031857.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 NM_025337.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 AK002664.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 AK003915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 AK143203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 BC031857.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P188795 NM_025337.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 AK002664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 AK003915.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 AK143203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 BC031857.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 NM_025337.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 AK002664.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 AK003915.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 AK143203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 BC031857.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P188795 NM_025337.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23351049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05424738 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164675 - -
          Whole Mouse Genome G4122A A_51_P188795 AK002664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 AK003915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 AK143203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 BC031857.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 NM_025337.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 AK002664.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 AK003915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 AK143203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 BC031857.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P188795 NM_025337.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123778 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03481806 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03481822 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03481804 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351041 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351043 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23351040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23351106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04053527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03481810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04053532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03481815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23351059 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509640 - -
          Affymetrix HT MG-430 PM Array Plate 1458300_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417294_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438315_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509635 - -
          Mouse Genome 430 2.0 Array 1417294_at - -
          Mouse Genome 430 2.0 Array 1458300_PM_at - -
          Mouse Genome 430 2.0 Array 1417294_PM_at - -
          Mouse Genome 430 2.0 Array 1438315_x_at - -
          Mouse Genome 430 2.0 Array 1438315_PM_x_at - -
          Mouse Genome 430 2.0 Array 1458300_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110198_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110198_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400014262.mm.1 NM_025337.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014262.mm.1 NM_025337.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014262.mm.1 TC0400001743.mm.1 MTA TC
          Mouse Expression Set 430 1438315_x_at - -
          Mouse Expression Set 430 1417294_at - -
          Mouse Expression Set 430 1417294_PM_at - -
          Mouse Expression Set 430 1438315_PM_x_at - -
          Mouse Expression Set 430 1458300_PM_at - -
          Mouse Expression Set 430 1458300_at - -
          Murine Genome U74v2 Set 96069_at - -
          Murine Genome U74v2 Set 96069_PM_at - -
          Mouse Genome 430A 2.0 Array 1438315_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1417294_PM_at - -
          Mouse Genome 430A 2.0 Array 1438315_x_at - -
          Mouse Genome 430A 2.0 Array 1417294_at - -
          Affymetrix HT Mouse Genome 430A Array 1417294_at - -
          Affymetrix HT Mouse Genome 430A Array 1417294_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1438315_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438315_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1458300_at - -
          Affymetrix HT Mouse Genome 430B Array 1458300_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910142 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460672 - -
          MouseWG-6 v2 Expression BeadChip 2260341 - -
          Illumina mouse-6 v1.0 expression beadchip 460672 - -
          Illumina mouse-6 v1.0 expression beadchip 2260341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256518 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625991 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256518 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625991 - -
          mouse-6 v1.1 (Illumina) scl0002846.1_27-S - -
          mouse-6 v1.1 (Illumina) scl24777.7.1_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260341 - -
          Illumina Sentrix mouse-6.v1 bead array 460672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002846.1_27-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24777.7.1_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256518 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2908045 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256518 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2908045 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24777.7.1_203-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline