| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117989 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117996 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117990 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117991 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117994 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117995 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
AK006018.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
AK011817.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
BC024423.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
NM_025644.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
AK006018.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
AK011817.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
BC024423.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P69789 |
NM_025644.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P132846 |
AK002930.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P132846 |
AK011817.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P132846 |
NM_001164561.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P132846 |
NM_025644.4 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
492151 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
AK038117.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
AK076629.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
AK129008.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
AK155386.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
AK159301.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
BC029233.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
BC038644.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
BC057036.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554136 |
NM_175137.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
296563 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855084 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
826473 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
95442 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P102837 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114904 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
AK006018.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
AK011817.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
BC024423.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
NM_025644.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
AK006018.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
AK011817.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
BC024423.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P69789 |
NM_025644.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P132846 |
AK002930.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P132846 |
AK011817.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P132846 |
NM_001164561.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P132846 |
NM_025644.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P251465 |
AK006018.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P251465 |
AK011817.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P251465 |
BC024423.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P251465 |
NM_025644.4 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117986 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117976 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117988 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117984 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117993 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117989 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117991 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117972 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117968 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117982 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117995 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117963 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117980 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117979 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117996 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117965 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117967 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117975 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32117970 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P102837 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03598810 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114904 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P1196922 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P542005 |
AK002930.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P542005 |
AK011817.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P542005 |
BC024423.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P542005 |
NM_001164561.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P542005 |
NM_025644.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P251465 |
AK006018.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P251465 |
AK011817.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P251465 |
BC024423.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P251465 |
NM_025644.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
AK006018.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
AK011817.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
BC024423.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
NM_025644.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
AK006018.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
AK011817.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
BC024423.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P69789 |
NM_025644.4 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132846 |
AK002930.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132846 |
AK011817.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132846 |
NM_001164561.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132846 |
NM_025644.4 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P132722 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P102837 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114904 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14374927 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14374890 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14374914 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14374923 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117990 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117994 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117996 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117989 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117991 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P32117995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117996 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117961 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117968 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117993 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117967 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117939 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117956 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117958 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117934 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117916 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117970 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117947 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117937 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117936 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117989 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117924 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117938 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117935 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117982 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117963 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117986 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117892 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117984 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117897 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117973 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32117880 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117968 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117953 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14396721 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117977 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117963 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117988 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117976 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14401731 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117972 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117986 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117984 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117979 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117995 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117980 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117961 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117989 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117954 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117965 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117956 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14374927 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117975 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14374916 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117966 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117993 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117958 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P32117959 |
- |
- |