| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959872 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959878 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959875 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959870 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959873 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959868 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959871 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959879 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959876 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959877 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
NM_026502.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
AK003413.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
AK018510.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
AK045905.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
BC019533.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
NM_026502.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
AK003413.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
AK018510.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
AK045905.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P196395 |
BC019533.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
16250 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
105462 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490655 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
503808 |
AK033481.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
503808 |
NM_177802.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
779474 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
962069 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
156038 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P125053 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
NM_026502.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
AK003413.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
AK018510.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
AK045905.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
BC019533.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
NM_026502.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
AK003413.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
AK018510.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
AK045905.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P196395 |
BC019533.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959873 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959897 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959885 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959906 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959867 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959875 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959901 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959884 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959883 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959899 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959886 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959909 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959870 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959890 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959888 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959911 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959891 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959910 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959866 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959880 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959913 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959879 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959871 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959914 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959907 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959905 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959916 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959893 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959878 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959895 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959898 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959882 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959887 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30959903 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08681545 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03254978 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P125053 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P196395 |
NM_026502.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
AK003413.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
AK018510.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
AK045905.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
BC019533.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
NM_026502.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
AK003413.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
AK018510.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
AK045905.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P196395 |
BC019533.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P140351 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P125053 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12824828 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12824825 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12824826 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12824824 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959868 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959871 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959879 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959876 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959877 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959870 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959873 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959872 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959875 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30959878 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959776 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959903 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959916 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959942 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959873 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959847 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959832 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959871 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959843 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959784 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959758 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959875 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959779 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959828 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959897 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959810 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959841 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959893 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959878 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959945 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959812 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959888 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959867 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959855 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959919 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959830 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959820 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959775 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959880 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959864 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959926 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959821 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959763 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959803 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959766 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959840 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959760 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959824 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959861 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959838 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959946 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959952 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959767 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959943 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959881 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959800 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959788 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959906 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959787 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959939 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959836 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959791 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959822 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959890 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959918 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959845 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959853 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959819 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959857 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959851 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959772 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959913 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30959808 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959913 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959911 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959787 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959935 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959929 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959795 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12858575 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959887 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959791 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959792 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959909 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12865163 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959858 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12858584 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959891 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959865 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959760 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959866 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12594283 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959875 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12865161 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959784 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959885 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959901 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959907 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959916 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959903 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959759 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959851 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12605243 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959853 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959882 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959793 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959781 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959899 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12578184 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959867 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959832 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959834 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959931 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959919 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12531221 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959779 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959895 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959878 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959836 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12824831 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959830 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959848 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959925 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959857 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12824824 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959778 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959855 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959926 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959762 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12824828 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959847 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959898 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12684519 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959918 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12658347 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959757 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959920 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959845 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959897 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12630705 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959838 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12865160 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959933 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959881 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959843 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959871 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959786 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959914 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959864 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959788 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30959828 |
- |
- |