Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459514_m1
          See other IGSF6 GE Assays ›
          Gene Symbol:
          Igsf6
          Gene Name:
          immunoglobulin superfamily, member 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 121064067 - 121074903 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459514_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igsf6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030691.1 NP_109616.1 5 - 6 684 104
          GenBank mRNA AJ278575.1 - 5 - 6 684 104
          AK040569.1 - 5 - 6 665 104
          AK151417.1 - 5 - 6 673 104
          AK152186.1 - 5 - 6 673 104
          AK156382.1 - 5 - 6 622 104
          AK163058.1 - 5 - 6 665 104
          BC057951.1 - 5 - 6 681 5341788 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004207.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igsf6

          Panther Classification:

          Molecular Function -

          immunoglobulin

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK163058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 BC057951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK152186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK152580.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK156382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK163058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 BC057951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK152186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK152580.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P258372 AK156382.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114065 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126823 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK163058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 BC057951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK152186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK152580.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK156382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK163058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 BC057951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK152186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK152580.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P258372 AK156382.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25471978 - -
          Whole Mouse Genome G4122A A_51_P258372 AK163058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 BC057951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK152186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK152580.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK156382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK163058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 BC057951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK152186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK152580.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P258372 AK156382.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05999512 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06627491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05999507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06627500 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05999514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25472014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25471942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06627500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06505146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06621347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06621354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06396453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06049910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06332930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05955004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05999512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06505144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06627511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25471930 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10567583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567585 - -
          Affymetrix HT MG-430 PM Array Plate 1421408_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567580 - -
          Mouse Genome 430 2.0 Array 1421408_at - -
          Mouse Genome 430 2.0 Array 1421408_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80719_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80719_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700035031.mm.1 NM_030691.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035031.mm.1 NM_030691.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035031.mm.1 TC0700004207.mm.1 MTA TC
          Mouse Expression Set 430 1421408_PM_at - -
          Mouse Expression Set 430 1421408_at - -
          Mouse Genome 430A 2.0 Array 1421408_PM_at - -
          Mouse Genome 430A 2.0 Array 1421408_at - -
          Affymetrix HT Mouse Genome 430A Array 1421408_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421408_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5340348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715673 - -
          mouse-6 v1.1 (Illumina) scl30727.6.1_169-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30727.6.1_169-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2715673 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2715673 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline