Search
Search

Tmem2| Gene Symbol: Tmem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033759.2 | NP_001028931.1 | 12 - 13 | 2726 | 67 | |
| NM_031997.4 | NP_114386.3 | 13 - 14 | 2808 | 67 | ||
| GenBank mRNA | AK129352.1 | - | 7 - 8 | 1027 | 67 | |
| BC019745.1 | - | 4 - 5 | 449 | 3675225 | 67 | |
| BC076570.1 | - | 12 - 13 | 2332 | 6830791 | 67 | |
| BC089353.1 | - | 12 - 13 | 2708 | 6410437 | 67 | |
| BU058172.1 | - | 3 - 4 | 240 | 67 | ||
| CA752446.1 | - | 5 - 6 | 666 | 67 | ||
Set Membership: |
|