Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459644_m1
          See other LY6G6D GE Assays ›
          Gene Symbol:
          Ly6g6d
          Gene Name:
          lymphocyte antigen 6 complex, locus G6D
          Gene Aliases:
          A930024F17Rik, G6d, G6f, MEGT1, NG25, NG32
          Chromosome Location:
          Chr.17: 35071342 - 35080567 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459644_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly6g6d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033478.2 NP_258439.1 2 - 3 194 57
          GenBank mRNA AJ315548.1 - 2 - 3 194 57
          AJ315549.1 - 1 - 2 291 57
          BC131988.1 - 2 - 3 230 40130265 57
          BC131990.1 - 2 - 3 230 40130267 57
          BM933266.1 - 2 - 3 382 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001943.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly6g6d

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052145 BB737333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052145 NM_033478.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203196 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052145 BB737333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052145 NM_033478.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157416 - -
          Whole Mouse Genome G4122A A_52_P12590 BB737333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P12590 AJ315548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P12590 AJ315549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P12590 BC131988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P12590 BC131990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404246 BB737333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404246 AJ315548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404246 AJ315549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404246 BC131988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404246 BC131990.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13141025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13278720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13141023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13141031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13435668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157409 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157416 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450419 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450422 - -
          Affymetrix HT MG-430 PM Array Plate 1450774_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450418 - -
          Mouse Genome 430 2.0 Array 1450774_at - -
          Mouse Genome 430 2.0 Array 1450774_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114654_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114654_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700018450.mm.1 NM_033478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018450.mm.1 NM_033478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018450.mm.1 TC1700001943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018448.mm.1 NM_033478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018448.mm.1 NM_033478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018448.mm.1 TC1700001943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018449.mm.1 NM_033478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018449.mm.1 NM_033478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018449.mm.1 TC1700001943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018452.mm.1 NM_033478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018452.mm.1 NM_033478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018452.mm.1 TC1700001943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018446.mm.1 NM_033478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018446.mm.1 NM_033478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018446.mm.1 TC1700001943.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018447.mm.1 NM_033478.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018447.mm.1 NM_033478.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018447.mm.1 TC1700001943.mm.1 MTA TC
          Mouse Expression Set 430 1450774_PM_at - -
          Mouse Expression Set 430 1450774_at - -
          Murine Genome U74v2 Set 98286_PM_at - -
          Murine Genome U74v2 Set 98286_at - -
          Mouse Genome 430A 2.0 Array 1450774_PM_at - -
          Mouse Genome 430A 2.0 Array 1450774_at - -
          Affymetrix HT Mouse Genome 430A Array 1450774_at - -
          Affymetrix HT Mouse Genome 430A Array 1450774_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 361109 - -
          ABI Mouse Genome Survey Microarray 910186 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6770095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599251 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599251 - -
          mouse-6 v1.1 (Illumina) scl0114654.1_118-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0114654.1_118-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599251 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599251 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline