Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00459773_m1
          See other KRT82 GE Assays ›
          Gene Symbol:
          Krt82
          Gene Name:
          keratin 82
          Gene Aliases:
          Krt2-20
          Chromosome Location:
          Chr.15: 101541216 - 101550659 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00459773_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053249.3 NP_444479.2 2 - 3 673 58
          GenBank mRNA AY028606.1 - 2 - 3 681 58
          BC108971.2 - 2 - 3 673 40047321 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002262.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt82

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          structural molecule activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145626 AY028606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145626 NM_053249.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145626 AA607752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2145626 AF312019.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137310 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145626 AY028606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145626 NM_053249.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145626 AA607752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2145626 AF312019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239367 AY028606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239367 NM_053249.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239367 AA607752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239367 AF312019.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30492624 - -
          Whole Mouse Genome G4122A A_51_P239367 AY028606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239367 NM_053249.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239367 AA607752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239367 AF312019.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362014 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362028 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12356312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12356304 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362027 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12362024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30492588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12410984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12362024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12410990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11864960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12362014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11864955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12362027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12311286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12410995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12248924 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30492640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12042329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432733 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432732 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432726 - -
          Affymetrix Mouse Gene 1.1 ST Array 10432723 - -
          Mouse Genome 430 2.0 Array 1427719_s_at - -
          Mouse Genome 430 2.0 Array 1427719_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114566_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114566_at - -
          Affymetrix Mouse Clariom S Array PSR1500018606.mm.1 NM_053249.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018606.mm.1 NM_053249.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018606.mm.1 TC1500002262.mm.1 MTA TC
          Mouse Expression Set 430 1427719_PM_s_at - -
          Mouse Expression Set 430 1427719_s_at - -
          Mouse Genome 430A 2.0 Array 1427719_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427719_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 722992 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6370156 - -
          MouseWG-6 v2 Expression BeadChip 2350377 - -
          Illumina mouse-6 v1.0 expression beadchip 2350377 - -
          Illumina mouse-6 v1.0 expression beadchip 6370156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2604475 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227153 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2604475 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227153 - -
          mouse-6 v1.1 (Illumina) scl0064819.1_114-S - -
          mouse-6 v1.1 (Illumina) scl46707.9.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370156 - -
          Illumina Sentrix mouse-6.v1 bead array 2350377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46707.9.1_83-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0064819.1_114-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370156 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2927369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227153 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2604475 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2927369 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2604475 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227153 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline