Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460245_m1
          See other DNAJC9 GE Assays ›
          Gene Symbol:
          Dnajc9
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 9
          Gene Aliases:
          5330419I01Rik, AU020082
          Chromosome Location:
          Chr.14: 20384638 - 20388910 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460245_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134081.5 NP_598842.1 3 - 4 700 79
          GenBank mRNA AK075979.1 - 3 - 4 661 79
          AK077324.1 - 3 - 4 670 79
          AK153033.1 - 3 - 4 660 79
          AK162075.1 - 3 - 4 575 79
          BC014686.1 - 3 - 4 662 4481661 79
          BC023787.1 - 3 - 4 658 5346759 79
          BC027012.1 - 3 - 4 653 5367851 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001533.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajc9

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446597 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 BC023787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 BC027012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 BC014686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 AK075979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 AK077324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 BC023787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 BC027012.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 BC014686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 AK075979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162471 AK077324.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184224 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158281 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 BC023787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 BC027012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 BC014686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 AK075979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 AK077324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 BC023787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 BC027012.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 BC014686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 AK075979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162471 AK077324.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29446588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158281 - -
          Whole Mouse Genome G4122A A_51_P162471 BC023787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 BC027012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 BC014686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 AK075979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 AK077324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 BC023787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 BC027012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 BC014686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 AK075979.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162471 AK077324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446171 BC023787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446171 BC027012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446171 BC014686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446171 AK043725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P446171 AK075979.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11594452 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11626064 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11626061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11594446 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29446594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29446646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11626059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11731719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11731723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11359810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11594451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11677556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29446609 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10417833 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10417830 - -
          Affymetrix HT MG-430 PM Array Plate 1426473_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10417829 - -
          Mouse Genome 430 2.0 Array 1426473_at - -
          Mouse Genome 430 2.0 Array 1426473_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108671_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 108671_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400011413.mm.1 NM_134081.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011413.mm.1 NM_134081.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011413.mm.1 TC1400001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011412.mm.1 NM_134081.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011412.mm.1 NM_134081.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011412.mm.1 TC1400001533.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400011410.mm.1 NM_134081.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011410.mm.1 NM_134081.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400011410.mm.1 TC1400001533.mm.1 MTA TC
          Mouse Expression Set 430 1426473_PM_at - -
          Mouse Expression Set 430 1426473_at - -
          Murine Genome U74v2 Set 160203_PM_at - -
          Murine Genome U74v2 Set 160203_at - -
          Mouse Genome 430A 2.0 Array 1426473_at - -
          Mouse Genome 430A 2.0 Array 1426473_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426473_at - -
          Affymetrix HT Mouse Genome 430A Array 1426473_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 881197 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102230452 - -
          Illumina mouse-6 v1.0 expression beadchip 6130019 - -
          Illumina mouse-6 v1.0 expression beadchip 102230452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130019 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224989 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2576142 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224989 - -
          mouse-6 v1.1 (Illumina) scl45857.5_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 102230452 - -
          Illumina Sentrix mouse-6.v1 bead array 6130019 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45857.5_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130019 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102230452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224989 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224989 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2576142 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline