| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900001047.mm.1 |
NM_134154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001047.mm.1 |
NM_134154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001047.mm.1 |
TC1900000090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001049.mm.1 |
NM_134154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001049.mm.1 |
NM_134154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001049.mm.1 |
TC1900000090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001046.mm.1 |
NM_134154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001046.mm.1 |
NM_134154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001046.mm.1 |
TC1900000090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001048.mm.1 |
NM_134154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001048.mm.1 |
NM_134154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001048.mm.1 |
TC1900000090.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900001044.mm.1 |
NM_134154.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001044.mm.1 |
NM_134154.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900001044.mm.1 |
TC1900000090.mm.1 |
MTA TC |