Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460530_m1
          See other CATSPER1 GE Assays ›
          Gene Symbol:
          Catsper1
          Gene Name:
          cation channel, sperm associated 1
          Gene Aliases:
          Catsper, KSper
          Chromosome Location:
          Chr.19: 5335602 - 5344281 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460530_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Catsper1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139301.2 NP_647462.1 10 - 11 1919 74
          GenBank mRNA AF407332.1 - 10 - 11 1919 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000061.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Catsper1

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          calcium ion transport
          multicellular organism development
          spermatogenesis
          fusion of sperm to egg plasma membrane
          cell differentiation
          sperm motility
          regulation of ion transmembrane transport
          regulation of calcium ion transport
          transmembrane transport
          regulation of cilium beat frequency involved in ciliary motility
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          ion channel activity
          calcium activated cation channel activity
          voltage-gated ion channel activity
          voltage-gated calcium channel activity
          calcium channel activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1087486 NM_139301.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1087486 AF407332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1087486 NM_139301.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1087486 AF407332.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 304639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686787 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130681 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1087486 NM_139301.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1087486 AF407332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1087486 NM_139301.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1087486 AF407332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P220848 AF407332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1087486 NM_139301.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1087486 AF407332.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1087486 NM_139301.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P1087486 AF407332.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31928072 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130681 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09093089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09093104 - -
          Whole Mouse Genome G4122A A_52_P1087486 NM_139301.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1087486 AF407332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1087486 NM_139301.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1087486 AF407332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220848 AF407332.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113354 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136029 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130681 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14155359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14292473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14292474 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14162255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31928208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14498629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14319348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14498635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14155343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14167840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14425811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14155359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14292473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14480017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14155349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14424648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14167838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31928138 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10460572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460564 - -
          Affymetrix HT MG-430 PM Array Plate 1426281_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10460560 - -
          Mouse Genome 430 2.0 Array 1426281_PM_at - -
          Mouse Genome 430 2.0 Array 1426281_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225865_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225865_at - -
          Affymetrix Mouse Clariom S Array PSR1900000762.mm.1 NM_139301.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000762.mm.1 NM_139301.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000762.mm.1 TC1900000061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000763.mm.1 NM_139301.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000763.mm.1 NM_139301.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000763.mm.1 TC1900000061.mm.1 MTA TC
          Mouse Expression Set 430 1426281_PM_at - -
          Mouse Expression Set 430 1426281_at - -
          Murine Genome U74v2 Set 166358_at - -
          Murine Genome U74v2 Set 166358_PM_at - -
          Mouse Genome 430A 2.0 Array 1426281_at - -
          Mouse Genome 430A 2.0 Array 1426281_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426281_at - -
          Affymetrix HT Mouse Genome 430A Array 1426281_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 336784 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1240301 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001240301 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2617459 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2617459 - -
          mouse-6 v1.1 (Illumina) scl52834.10.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 1240301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52834.10.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240301 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2617459 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2617459 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline